Skip to Content
Merck
All Photos(1)

Key Documents

M1567

Sigma-Aldrich

Malic Dehydrogenase from porcine heart

≥600 units/mg protein (biuret), ammonium sulfate suspension

Synonym(s):

L-Malate: NAD+ oxidoreductase, MDH, Malate Dehydrogenase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

form

ammonium sulfate suspension

Quality Level

specific activity

≥600 units/mg protein (biuret)

foreign activity

Glutamic-Oxalacetic Transaminase ≤0.01%
Glutamic-Pyruvic Transaminase ≤0.01%

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Malic Dehydrogenase is a ubiquitous enzyme, which exists in two isoforms in eukaryotic cells.[1]
Malic dehydrogenase exists as a dimer with each subunit containing an NAD-binding domain and a substrate-binding carboxy-terminal domain required for activity. [2]
Malic dehydrogenase is a cytoplasmic isozyme and an important catalyst in the tricarboxylic acid cycle.

Application

Malic Dehydrogenase from porcine heart has been used:
  • in qualitative protein binding measurements[3]
  • to test internally calibrated electrochemical continuous enzyme assay (ICECEA) with model enzyme pair[4]
  • to investigate the effect of chaperone on the refolding of heat-denatured malate dehydrogenase[5]

Malic dehydrogenase has been used in a study to assess the effect of an immunomodulator S2 complex on the enzymes of the parasites. [6] It has also been used in a study to investigate the heterogeneity of lactic and malic dehydrogenase in cerebrospinal fluid. [7]

Biochem/physiol Actions

Malic Dehydrogenase (MDH) plays an important role in the citric acid cycle in mitochondria. It catalyzes the interconversion of substrates malate and oxaloacetate with the simultaneous oxidation/reduction of NAD/NADH+. MDH present in the cytosol is involved in the shuttling of malate/aspartate.[1]

Unit Definition

One unit will convert 1.0 μmole of oxalacetate and β-NADH to L-malate and β-NAD per min at pH 7.5 at 25°C.

Physical form

Suspension in 2.8 M (NH4)2SO4 solution, pH 6.0

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical Coupled-Enzyme Assays at Carbon Nanotubes
Zhang M, et al.
Analytical Chemistry, 86(18), 9330-9334 (2014)
Heterogeneity of lactic and malic dehydrogenase in cerebrospinal fluid
Lowenthal, A., et al.
Journal of Neurochemistry, 7, 135-140 (2006)
Malate dehydrogenases-structure and function
Minarik P, et al.
General Physiology and Biophysics, 21(3), 257-266 (2002)
Sensing NADH conformation using phasor analysis on fluorescence spectra
Palo D, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 186(3), 105-111 (2017)
Sandip K Nandi et al.
Biochemical and biophysical research communications, 533(4), 1352-1358 (2020-10-22)
The chaperone activity of α-crystallin is important for maintaining the transparency of the human lens. αB-crystallin (αBC) is a long-lived protein in the lens that accumulates chemical modifications during aging. The formation of advanced glycation end products (AGEs) through glycation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service