Skip to Content
Merck
All Photos(2)

Documents

M6680

Sigma-Aldrich

Se-(Methyl)selenocysteine hydrochloride

≥95% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NO2Se · HCl
CAS Number:
Molecular Weight:
218.54
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

Assay

≥95% (TLC)

storage temp.

−20°C

SMILES string

Cl.C[Se]C[C@H](N)C(O)=O

InChI

1S/C4H9NO2Se.ClH/c1-8-2-3(5)4(6)7;/h3H,2,5H2,1H3,(H,6,7);1H/t3-;/m0./s1

InChI key

JMPVTFHGWJDSDV-DFWYDOINSA-N

General description

Selenium (Se)-(Methyl) selenocysteine (MeSeCys) is a methylated product of selenocysteine, this reaction is catalyzed by selenocysteine methyltransferase (SMT) in Astragalus bisulcatus.

Application

Se-(Methyl) selenocysteine hydrochloride has been used as a standard in fluorescence-XANES (X-ray absorption near-edge spectroscopy) imaging.

Biochem/physiol Actions

Selenium (Se)-(Methyl) selenocysteine functions as a potent chemotherapeutic agent for human breast cancer. Enhanced production of methlylselenol from L-Se-methylselenocysteine in vivo alters redox status of cells and oxidatively triggers cellular apoptosis. It is implicated in inhibition of mammary tumor formation in rats.
Se-(Methyl)selenocysteine is a chemopreventive agent that blocks cell cycle progression and proliferation of premalignant mammary lesions and induces apoptosis of cancer cell lines in culture.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In situ speciation and distribution of toxic selenium in hydrated roots of cowpea.
Wang P
Plant Physiology, 163(1), 407-418 (2013)
Production of Se-methylselenocysteine in transgenic plants expressing selenocysteine methyltransferase.
Ellis DR
BMC plant biology, 4:1 (2004)
Se-methylselenocysteine: a new compound for chemoprevention of breast cancer.
Medina D
Nutrition and Cancer, 40(1), 12-17 (2001)
N V Dolgova et al.
Metallomics : integrated biometal science, 8(3), 305-312 (2016-01-20)
Selenium is an essential micronutrient for many organisms, and in vertebrates has a variety of roles associated with protection from reactive oxygen species. Over the past two decades there have been conflicting reports upon human health benefits and detriments arising
Dimethyldiselenide and methylseleninic acid generate superoxide in an in vitro chemiluminescence assay in the presence of glutathione: implications for the anticarcinogenic activity of L-selenomethionine and L-Se-methylselenocysteine.
Spallholz JE
Nutrition and Cancer, 40(1), 34-41 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service