Skip to Content
Merck
All Photos(1)

Key Documents

S3875

Sigma-Aldrich

Seleno-DL-methionine

≥99% (TLC)

Synonym(s):

2-amino-4-(methylseleno)-Butanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO2Se
CAS Number:
Molecular Weight:
196.11
Beilstein:
1758204
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

Seleno-DL-methionine, ≥99% (TLC)

Assay

≥99% (TLC)

form

powder

color

white to off-white

application(s)

cell analysis

storage temp.

−20°C

SMILES string

C[Se]CCC(N)C(O)=O

InChI

1S/C5H11NO2Se/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)

InChI key

RJFAYQIBOAGBLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephanie A Schaefer et al.
Journal of molecular biology, 425(2), 222-231 (2012-11-20)
Sulfur, a key contributor to biological reactivity, is not amendable to investigations by biological NMR spectroscopy. To utilize selenium as a surrogate, we have developed a generally applicable (77)Se isotopic enrichment method for heterologous proteins expressed in Escherichia coli. We
Pedro José Barbosa Pereira et al.
Journal of structural biology, 181(2), 89-94 (2012-11-28)
Selenomethionine labeling is the most common technique used in protein crystallography to derivatize recombinant proteins for experimental phasing using anomalous scattering at tunable synchrotron beamlines. Recently, it has been shown that UV radiation depletes electron density of selenium atoms of
Sergio Trillo-Muyo et al.
Acta crystallographica. Section D, Biological crystallography, 69(Pt 3), 464-470 (2013-03-23)
While small organic molecules generally crystallize forming tightly packed lattices with little solvent content, proteins form air-sensitive high-solvent-content crystals. Here, the crystallization and full structure analysis of a novel recombinant 10 kDa protein corresponding to the C-terminal domain of a
Jiankun Duan et al.
Journal of chromatography. A, 1268, 173-179 (2012-11-13)
A new phenylalanine derivative (L-N-(2-hydroxy-propyl)-phenylalanine, L-HP-Phe) was synthesized and its chelate with Cu(II) (Cu(II)-(L-HP-Phe)(2)) was used as the chiral selector for the ligand-exchange (LE) chiral separation of D,L-selenomethionine (SeMet) in selenized yeast samples by micelle electrokinetic capillary chromatography (MEKC). In
Qun Liu et al.
Science (New York, N.Y.), 336(6084), 1033-1037 (2012-05-26)
Crystal structure analyses for biological macromolecules without known structural relatives entail solving the crystallographic phase problem. Typical de novo phase evaluations depend on incorporating heavier atoms than those found natively; most commonly, multi- or single-wavelength anomalous diffraction (MAD or SAD)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service