Skip to Content
Merck
All Photos(1)

Documents

41572

Sigma-Aldrich

Dimethyl selenide

≥99.0% (GC)

Synonym(s):

Methyl selenide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2Se
CAS Number:
Molecular Weight:
109.03
Beilstein:
1696848
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.0% (GC)

form

liquid

density

1.408 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C[Se]C

InChI

1S/C2H6Se/c1-3-2/h1-2H3

InChI key

RVIXKDRPFPUUOO-UHFFFAOYSA-N

General description

Atomic number of base material: 34 Selenium

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ximena Diaz et al.
Environmental science & technology, 43(1), 53-59 (2009-02-13)
The removal mechanisms that govern Se concentrations in the Great Salt Lake are unknown despite this terminal lake being an avian habitat of hemispheric importance. However, the volatilization flux of Se from the Great Salt Lake has not been previously
Mikhail V Andreev et al.
Molecules (Basel, Switzerland), 25(24) (2020-12-19)
The efficient regio- and stereoselective synthesis of (Z,Z)-3,3'-selanediylbis(2-propenamides) in 76-93% yields was developed based on the reaction of sodium selenide with 3-trimethylsilyl-2-propynamides. (Z,Z)-3,3'-Selanediylbis(2-propenamides) are a novel class of organoselenium compounds. To date, not a single representative of 3,3'-selanediylbis(2-propenamides) has been
Daniel Kremer et al.
Analytical and bioanalytical chemistry, 383(3), 509-515 (2005-09-15)
The amount of volatile dimethylselenide (DMSe) in breath has been monitored after ingestion of sub-toxic amounts of selenium (300 microg (77)Se, as selenite) by a healthy male volunteer. The breath samples were collected in Tedlar bags every hour in the
Yuki Ohta et al.
Chemical research in toxicology, 22(11), 1795-1801 (2009-09-01)
Selenium is an essential trace element found in vegetables as selenomethionine (SeMet) and methylselenocysteine (MeSeCys). In the present study, we used stable isotopes of Se to investigate differences between how SeMet and MeSeCys are metabolized, using methylseleninic acid (MSA) as
Dijana Juresa et al.
Analytical chemistry, 78(24), 8569-8574 (2006-12-15)
Recent work has shown the presence of volatile selenium metabolites in human urine and suggested that these compounds could compromise quantitative selenium analyses by ICPMS. We show that with a commonly used sample introduction system (pneumatic nebulizer and spray chamber)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service