Skip to Content
Merck
All Photos(1)

Key Documents

M6413

Sigma-Aldrich

L-(−)-Malic acid

BioXtra, ≥95%

Synonym(s):

(S)-(−)-2-Hydroxysuccinic acid, L-Hydroxybutanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH(OH)CO2H
CAS Number:
Molecular Weight:
134.09
Beilstein:
1723541
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

grade

BioPerformance Certified

product line

BioXtra

Assay

≥95%

form

powder

impurities

≤0.01% Phosphorus (P)
≤0.1% Insoluble matter

ash

<0.1%

mp

101-103 °C (lit.)

solubility

H2O: 0.5 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

O[C@@H](CC(O)=O)C(O)=O

InChI

1S/C4H6O5/c5-2(4(8)9)1-3(6)7/h2,5H,1H2,(H,6,7)(H,8,9)/t2-/m0/s1

InChI key

BJEPYKJPYRNKOW-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margrethe A Olesen et al.
Redox biology, 34, 101558-101558 (2020-05-25)
Aging is a process characterized by cognitive impairment and mitochondrial dysfunction. In neurons, these organelles are classified as synaptic and non-synaptic mitochondria depending on their localization. Interestingly, synaptic mitochondria from the cerebral cortex accumulate more damage and are more sensitive
Chao Yang et al.
Cell host & microbe, 27(3), 467-475 (2020-02-23)
Fecal IgA production depends on colonization by a gut microbiota. However, the bacterial strains that drive gut IgA production remain largely unknown. Here, we assessed the IgA-inducing capacity of a diverse set of human gut microbial strains by monocolonizing mice
Jennifer Ngo et al.
STAR protocols, 2(1), 100243-100243 (2021-01-19)
Mitochondria play a central role in lipid metabolism and can bind to lipid droplets. However, the role and functional specialization of the population of peridroplet mitochondria (PDMs) remain unclear, as methods to isolate functional PDMs were not developed until recently.
Nikhil Madhusudhan et al.
ACS chemical biology, 15(1), 158-170 (2019-12-25)
Selective toxicity among cancer cells of the same lineage is a hallmark of targeted therapies. As such, identifying compounds that impair proliferation of a subset of non-small-cell lung cancer (NSCLC) cell lines represents one strategy to discover new drugs for
Kaiyue Wu et al.
Biochemical and biophysical research communications, 525(3), 733-739 (2020-03-08)
Cigarette smoke is one of major risk factors in the pathogenesis of chronic obstructive pulmonary disease (COPD). It is generally believed that cigarette smoke induces mitochondrial damage in the alveolar epithelial cells to contribute to COPD. However, the exact molecular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service