Skip to Content
Merck
All Photos(4)

Key Documents

M1000

Sigma-Aldrich

L-(−)-Malic acid

≥95% (titration)

Synonym(s):

(S)-(−)-2-Hydroxysuccinic acid, L-Hydroxybutanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH(OH)CO2H
CAS Number:
Molecular Weight:
134.09
Beilstein:
1723541
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95% (titration)

form

powder

pKa (25 °C)

(1) 3.46, (2) 5.10

mp

101-103 °C ((lit.))
101-103 °C (lit.)

solubility

water: 100 mg/mL, clear to very slightly hazy, colorless

density

1,595  g/cm3 at 68 °F

SMILES string

O[C@@H](CC(O)=O)C(O)=O
O[C@@H](CC(O)=O)C(O)=O

InChI

1S/C4H6O5/c5-2(4(8)9)1-3(6)7/h2,5H,1H2,(H,6,7)(H,8,9)/t2-/m0/s1
1S/C4H6O5/c5-2(4(8)9)1-3(6)7/h2,5H,1H2,(H,6,7)(H,8,9)/t2-/m0/s1

InChI key

BJEPYKJPYRNKOW-REOHCLBHSA-N
BJEPYKJPYRNKOW-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Malic acid is the naturally occurring isomer of malic acid, found mainly in sour and unripe fruits.

Application

L-(-)-Malic acid may be used to prepare:
  • diethyl (S)-malate
  • ethyl (R)-2-hydroxyl-4-phenylbutanoate
  • ethyl (S)-2-hydroxyl-4-phenylbutanoate
  • D-homophenylalanine ethyl ester hydrochloride
  • furo[3,2-i]indolizines

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric synthesis of furo [3, 2-i] indolizines from L-malic acid.
Lee YS, et al.
Tetrahedron, 55(15), 4631-4636 (1999)
A practical synthesis of ethyl (R)-and (S)-2-hydroxy-4-phenylbutanoate and D-homophenylalanine ethyl ester hydrochloride from L-malic acid.
Lin WQ, et al.
Tetrahedron Asymmetry, 12(11), 1583-1587 (2001)
Lei Shi et al.
Science (New York, N.Y.), 368(6497) (2020-05-23)
Although perovskite solar cells have produced remarkable energy conversion efficiencies, they cannot become commercially viable without improvements in durability. We used gas chromatography-mass spectrometry (GC-MS) to reveal signature volatile products of the decomposition of organic hybrid perovskites under thermal stress.
A stereospecific synthesis of (-)-Bestatin from L-malic acid.
Norman BH and Morris ML.
Tetrahedron Letters, 33(45), 6803-6806 (1992)
Mc Murry JE
Organic Chemistry , Biological Approach (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service