Skip to Content
Merck
All Photos(2)

Key Documents

M3047

Sigma-Aldrich

Mizoribine

≥98% (TLC)

Synonym(s):

N′-(β-D-Ribofuranosyl)-5-hydroxy­imida­zole-4-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N3O6
CAS Number:
Molecular Weight:
259.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

storage temp.

2-8°C

SMILES string

NC(=O)c1ncn([C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)c1O

InChI

1S/C9H13N3O6/c10-7(16)4-8(17)12(2-11-4)9-6(15)5(14)3(1-13)18-9/h2-3,5-6,9,13-15,17H,1H2,(H2,10,16)/t3-,5-,6-,9-/m1/s1

InChI key

HZQDCMWJEBCWBR-UUOKFMHZSA-N

Application

Mizoribine has been used in topical treatment to evaluate its effect on ocular surface damage of dry eye in B6 mice subjected to desiccating stress (DS). It has also been used as an inosine monophosphate dehydrogenase (IMPDH) inhibitor to test its in vivo efficacy.

Biochem/physiol Actions

Mizoribine is an imidazole nucleoside possessing strong immunosuppressive properties. It selectively blocks T-cell proliferation response to mitogenic and allo-antigenic stimulation. Mizoribine blocks the movement of T cells from G to S phase. In addition, it significantly decreases the number of B cells at the S, G, and M phases. Mizoribine inhibits de novo synthesis of nucleotides by inhibition of inosine monophosphate dehydrogenase. The resulting nucleotide depletion inhibits DNA synthesis.(3)

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kohei Miyazaki et al.
Clinical nephrology, 77(4), 261-266 (2012-03-27)
Imbalance between T-helper 1 (Th1) and 2 (Th2) lymphocytes and effects of reactive oxygen species (ROS) upon glomerular capillary walls have been implicated in minimal change nephrotic syndrome (MCNS). By polymerase chain reaction and comparative genomic hybridization, we evaluated mutations
Napoli, K.L.
J. Inter. Fed. Clin. Chem., 4, 15-15 (1992)
Long-term multidrug therapy in an adolescent patient with proliferative lupus nephritis: a trial of less cytotoxic therapy.
Tomomi Aizawa-Yashiro et al.
Clinical nephrology, 78(4), 332-334 (2012-08-03)
Topical Application of Mizoribine Suppresses CD4+ T-cell-Mediated Pathogenesis in Murine Dry Eye
Zhang X, et al.
Investigative Ophthalmology & Visual Science, 58(14), 6056-6064 (2017)
Kazuya Ishida et al.
Drug metabolism and pharmacokinetics, 28(1), 75-80 (2012-07-21)
Bioavailability of mizoribine in subjects with the concentrative nucleoside transporter 1 (CNT1, SLC28A1) 565-A/A allele is significantly lower than that in subjects with the SLC28A1 565-G/G allele. The aims of the present study were to investigate the cellular uptake of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service