Skip to Content
Merck
All Photos(2)

Key Documents

SML0737

Sigma-Aldrich

MitoTEMPO

≥98% (HPLC), powder, antioxidant

Synonym(s):

(2-(2,2,6,6-Tetramethylpiperidin-1-oxyl-4-ylamino)-2-oxoethyl)triphenylphosphonium chloride, Mito-TEMPO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H35N2O2P · Cl
CAS Number:
Molecular Weight:
510.03
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

MitoTEMPO, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated
protect from light

color

white to light brown

solubility

H2O: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

[P+](CC(=O)NC4CC(N(C(C4)(C)C)[O])(C)C)(c3ccccc3)(c2ccccc2)c1ccccc1.[Cl-]

InChI

1S/C29H34N2O2P.ClH/c1-28(2)20-23(21-29(3,4)31(28)33)30-27(32)22-34(24-14-8-5-9-15-24,25-16-10-6-11-17-25)26-18-12-7-13-19-26;/h5-19,23H,20-22H2,1-4H3;1H

InChI key

WKKFJIJNGHNQQW-UHFFFAOYSA-N

Application

MitoTEMPO has been used as a mitochondria-targeted antioxidant.

Biochem/physiol Actions

MitoTEMPO is a mitochondrially targeted antioxidant, a specific scavenger of mitochondrial superoxide.
MitoTEMPO is a mitochondrially targeted antioxidant, a specific scavenger of mitochondrial superoxide. MitoTEMPO is a combination of the antioxidant piperidine nitroxide TEMPO with the lipophilic cation triphenylphosphonium, giving MitoTEMPO the ability to pass through lipid bilayers with ease and accumulate several hundred-fold in mitochondria. Mitochondrial dysfunction and oxidative stress contribute to a variety of disorders involving cardiovascular, kidney, liver disorders, Parkinson′s and Alzheimer′s disease.

Other Notes

Light sensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eunice E To et al.
Antioxidants & redox signaling, 32(13), 929-942 (2019-06-14)
Aims: Reactive oxygen species (ROS) are highly reactive molecules generated in different subcellular sites or compartments, including endosomes via
Regulation of morphine-induced synaptic alterations: Role of oxidative stress, ER stress, and autophagy
Yu C, et al.
The Journal of Biological Chemistry (2016)
Caveolin 1-related autophagy initiated by aldosterone-induced oxidation promotes liver sinusoidal endothelial cells defenestration
Xiaoying L, et al.
Redox Biology, 508-521 (2017)
Modulation of mitochondrial antiviral signaling by human herpesvirus 8 interferon regulatory factor 1
Young KH, et al.
Journal of Virology (2016)
Zimei Wu et al.
Zebrafish, 16(2), 171-181 (2019-02-07)
Chemical interventions are regularly used to examine and manipulate macrophage function in larval zebrafish. Given chemicals are typically administered by simple immersion or injection, it is not possible to resolve whether their impact on macrophage function is direct or indirect.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service