Skip to Content
Merck
All Photos(1)

Key Documents

L9668

Sigma-Aldrich

Lidoflazine

≥98% (HPLC), powder

Synonym(s):

4-[4,4-Bis(4-fluorophenyl)butyl]-N-(2,6-dimethylphenyl)-1-piperazineacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H35F2N3O
CAS Number:
Molecular Weight:
491.62
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

Cc1cccc(C)c1NC(=O)CN2CCN(CCCC(c3ccc(F)cc3)c4ccc(F)cc4)CC2

InChI

1S/C30H35F2N3O/c1-22-5-3-6-23(2)30(22)33-29(36)21-35-19-17-34(18-20-35)16-4-7-28(24-8-12-26(31)13-9-24)25-10-14-27(32)15-11-25/h3,5-6,8-15,28H,4,7,16-21H2,1-2H3,(H,33,36)

InChI key

ZBIAKUMOEKILTF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Lidoflazine is an antianginal calcium channel blocker that carries a significant risk of QT interval prolongation and ventricular arrhythmia. It prolongs QT interval by blocking HERG channel. IC50 < 0.1 μM

Features and Benefits

This compound is featured on the Calcium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liver preservation with lidoflazine and the University of Wisconsin solution: a dose-finding study.
J Jacobsson et al.
Transplantation, 56(2), 472-475 (1993-08-01)
J Jacobsson et al.
Transplant international : official journal of the European Society for Organ Transplantation, 6(5), 281-284 (1993-01-01)
The effect of adding a 21-aminosteroid, U74500A, and a Ca2+ antagonist, lidoflazine, alone and together to UW solution was assessed in a rat liver preservation model. Following preservation, the livers were reperfused using a closed circuit, and the release of
G J Grover et al.
The Journal of pharmacology and experimental therapeutics, 268(1), 90-96 (1994-01-01)
The goal of this study was to determine the cardioprotective profile for the nucleoside transport inhibitor 2-(aminocarboxyl)-N-(4-amino-2,6-dichlorophenyl)-4-[5,5-bis(4- fluorophenyl)pentyl]-1-piperazinylacetamide trihydrochloride-2,5 hydrate (R 75231) in isolated rat hearts and whether its protective effects are caused by adenosine A1 activation. R 75231 increased
Thomas P Zonneveld et al.
The Cochrane database of systematic reviews, 7, CD007858-CD007858 (2018-07-20)
Stroke is an important cause of death and disability worldwide. Since high blood pressure is an important risk factor for stroke and stroke recurrence, drugs that lower blood pressure might play an important role in secondary stroke prevention. To investigate
S G De Hert et al.
Journal of cardiothoracic and vascular anesthesia, 11(1), 42-48 (1997-02-01)
The present study evaluated the effects of the nucleoside transport inhibitor, lidoflazine, at a dose of 1 mg/kg, on left ventricular function. Patients were randomly assigned to receive either lidoflazine or saline in a double-blind manner. A university hospital. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service