Recommended Products
Quality Level
Related Categories
General description
Glycan analysis has become an increasingly critical aspect of glycomics and proteomics, due to the role of glycoproteins in cell signaling, cell adhesion, immune response, and disease states. Since they tend to have low spectral activity in both UV and visible light, it is often necessary to label glycans in order to enhance detection. GlycoProfile™ Glycan Clean-Up Cartridges are designed for the post-labeling cleanup of labelled glycans when using the GlycoProfile™ 2-AA or 2-AB Labeling Kits. Glycans adsorb to the membrane while excess dye and other hydrophobic contaminants pass through. Mono- and disaccharides have a weak affinity for the cartridge membrane and tend to bind poorly. Purified glycans are subsequently desorbed and ready for further enzymatic digestion or analysis by a number of methods, including HPLC.
Application
GlycoProfile™ Glycan Clean-Up Cartridge is used for clean up of glycan samples after reductive amination labeling or enzymatic digestion. It has also been used to remove excess dye and hydrophobic contaminants from N-linked glycans. The cartridge is equilibrated with acetonitrile, then loaded with a maximum of 20 μg of a glycan sample. The glycans are bound while excess dye and salts are removed by washing with acetonitrile. The glycans are then eluted with water.
Linkage
Recommended for use with the GlycoProfile 2-AA (Cat. No. PP0530) and 2-AB (Cat. No. PP0520) Labeling Kits.
Storage and Stability
Store the cartridges at room temperature. The cartridges are stable for at least one year at room temperature, as supplied.
Other Notes
Each cartridge is recommended for single use only. The maximum sample size is 10 μL of glycan containing solution and/or 20 μg of glycan.
Legal Information
GlycoProfile is a trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.
GlycoProfile is a trademark of Sigma-Aldrich Co. LLC
Disclaimer
This product is for R&D use only, not for drug, household, or other uses. Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Substitution of glutamine by glutamate enhances production and galactosylation of recombinant IgG in Chinese hamster ovary cells
Hong JK, et al.
Applied Microbiology and Biotechnology, 88(4), 869-876 (2010)
Preclinical Characterization of CTL-1,
A Biosimilar Anti-EGFR Monoclonal
Antibody for Cetuximab
A Biosimilar Anti-EGFR Monoclonal
Antibody for Cetuximab
Hu P, et al.
Journal of Chemical and Pharmaceutical Sciences
, 1(1), 1003-1003 (2017)
Recent trends in analytical and structural glycobiology
Novotny MV, et al.
Current Opinion in Chemical Biology, 17(5), 832-840 (2013)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service