Skip to Content
Merck
All Photos(1)

Key Documents

F9427

Sigma-Aldrich

Fexofenadine hydrochloride

>98% (HPLC)

Synonym(s):

Fexofenidine hydrochloride, MDL 16455 hydrochloride, Terfenidine carboxylate hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H39NO4 · HCl
CAS Number:
Molecular Weight:
538.12
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

>98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: soluble ≥5 mg/mL

originator

Sanofi Aventis

storage temp.

−20°C

SMILES string

Cl[H].CC(C)(C(O)=O)c1ccc(cc1)C(O)CCCN2CCC(CC2)C(O)(c3ccccc3)c4ccccc4

InChI

1S/C32H39NO4.ClH/c1-31(2,30(35)36)25-17-15-24(16-18-25)29(34)14-9-21-33-22-19-28(20-23-33)32(37,26-10-5-3-6-11-26)27-12-7-4-8-13-27;/h3-8,10-13,15-18,28-29,34,37H,9,14,19-23H2,1-2H3,(H,35,36);1H

InChI key

RRJFVPUCXDGFJB-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

Application

Fexofenadine hydrochloride has been used as a non-cationic ampliphilic antihistamines in NCI-H661 cells and as a histamine H1 receptor antagonist in mice

Biochem/physiol Actions

Fexofenadine is a non-sedating H1 histamine receptor antagonist.
Fexofenadine is used in treating allergic rhinitis and chronic idiopathic urticaria. It may give relief in bothersome symptom and is a second-generation antihistamines. Fexofenadine provides better relief compared to other antihistamines for nasal congestion. It is the active metabolite of terfenadine and bring down allergen-induced symptoms.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmacokinetics of fexofenadine: evaluation of a microdose and assessment of absolute oral bioavailability
Lappin G, et al.
European Journal of Pharmaceutical Sciences, 40(2), 125-131 (2010)
Effect of the second-generation antihistamine, fexofenadine, on cough reflex sensitivity and pulmonary function
Dicpinigaitis PV and Gayle YE
British Journal of Clinical Pharmacology, 56(5), 501-504 (2003)
A review of the efficacy of desloratadine, fexofenadine, and levocetirizine in the treatment of nasal congestion in patients with allergic rhinitis
Bachert C
Clinical Therapeutics, 31(5), 921-944 (2009)
Inhibitory effect of oblongifolin C on allergic inflammation through the suppression of mast cell activation
Lu Y, et al.
Molecular and Cellular Biochemistry, 406(1-2), 263-271 (2015)
Jürgen Brüning et al.
The Journal of pharmacy and pharmacology, 67(6), 773-781 (2015-02-14)
This study describes the general method for the determination of the crystal structures of active pharmaceutical ingredients (API) from powder diffraction data and demonstrates its use to determine the hitherto unknown crystal structure of fexofenadine hydrochloride, a third-generation antihistamine drug.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service