Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lipopolysaccharides from
Escherichia coli
O127:B8
Synonym(s):
LPS
EC Number:
297-473-0
Products
Product Category
carbohydrates (5)
lipopolysaccarides (4)
Brand
Sigma-Aldrich (5)
Biological Source
Color
Physical Form
Product Line
Quality Segment
Sterilization Method
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
L3129
Lipopolysaccharides from
Escherichia coli
O127:B8,
purified by phenol extraction
Expand
View Pricing
L4516
Lipopolysaccharides from
Escherichia coli
O127:B8,
BioXtra, suitable for cell culture, γ-irradiated
Expand
View Pricing
L5024
Lipopolysaccharides from
Escherichia coli
O127:B8,
purified by ion-exchange chromatography, TLR ligand tested
Expand
View Pricing
L5668
Lipopolysaccharides from
Escherichia coli
O127:B8,
Ready Made solution, 1 mg/mL
Expand
View Pricing
L3137
Lipopolysaccharides from
Escherichia coli
O127:B8,
purified by gel-filtration chromatography
Expand
View Pricing