L3791
Lamotrigine
≥98% (TLC), powder, glutamate release inhibitor
Synonym(s):
6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine, GI 267119X
About This Item
Recommended Products
product name
Lamotrigine, ≥98%, powder
Quality Level
Assay
≥98%
form
powder
color
white
solubility
DMSO: soluble 20 mg/mL at 60 °C
H2O: insoluble
originator
GlaxoSmithKline
storage temp.
2-8°C
SMILES string
Nc1nnc(c(N)n1)-c2cccc(Cl)c2Cl
InChI
1S/C9H7Cl2N5/c10-5-3-1-2-4(6(5)11)7-8(12)14-9(13)16-15-7/h1-3H,(H4,12,13,14,16)
InChI key
PYZRQGJRPPTADH-UHFFFAOYSA-N
Gene Information
human ... SCN10A(6336) , SCN11A(11280) , SCN1A(6323) , SCN2A(6326) , SCN3A(6328) , SCN4A(6329) , SCN5A(6331) , SCN7A(6332) , SCN8A(6334) , SCN9A(6335)
rat ... Scn2a1(24766) , Scnn1a(25122)
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Features and Benefits
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service