Skip to Content
Merck
All Photos(1)

Key Documents

SML0248

Sigma-Aldrich

Zolmitriptan

≥98% (HPLC)

Synonym(s):

(4S)-4-((3-(2-(Dimethylamino)ethyl)-1H-indol-5-yl)methyl)-2-oxazolidinone, (S)-4-((3-(2-(Dimethylamino)ethyl)indol-5-yl)methyl)-2-oxazolidinone, 311C90

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H21N3O2
CAS Number:
Molecular Weight:
287.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -3 to -8°, c = 1 in methanol

color

white to beige

solubility

DMSO: ≥5 mg/mL

storage temp.

2-8°C

SMILES string

CN(C)CCc1c[nH]c2ccc(C[C@H]3COC(=O)N3)cc12

InChI

1S/C16H21N3O2/c1-19(2)6-5-12-9-17-15-4-3-11(8-14(12)15)7-13-10-21-16(20)18-13/h3-4,8-9,13,17H,5-7,10H2,1-2H3,(H,18,20)/t13-/m0/s1

InChI key

ULSDMUVEXKOYBU-ZDUSSCGKSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Zolmitriptan has been used as a 5-hydroxytryptamine receptor 1B (5-HT1B/D) activator to treat grafts to study its effects on the morphology of the ectopic eye.
Zolmitriptan has been used to induce hyper-innervation of transplants.

Biochem/physiol Actions

Zolmitriptan is a potent, selective. centrally and peripherally acting 5HT1B/1D receptor agonist.
Zolmitriptan is a selective serotonin receptor agonist of the 5HT1B and 5HT1D subtypes, both centrally and peripherally. It has been used clinically for the acute treatment of migraine attacks with or without aura and cluster headaches.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robhash Kusam Subedi et al.
International journal of pharmaceutics, 419(1-2), 209-214 (2011-08-17)
The effects of different formulation variables including pressure sensitive adhesive (PSA), thickness of the matrix, solvent system, inclusion of crystallization inhibitor, loading amount of drug and enhancers on the transdermal absorption of zolmitriptan were investigated. Acrylic adhesive with hydroxyl functional
Jessica M D'Amico et al.
Journal of neurophysiology, 109(6), 1485-1493 (2012-12-12)
Activation of receptors by serotonin (5-HT1) and norepinephrine (α2) on primary afferent terminals and excitatory interneurons reduces transmission in spinal sensory pathways. Loss or reduction of descending sources of serotonin and norepinephrine after spinal cord injury (SCI) and the subsequent
Tony W Ho et al.
Headache, 51(1), 64-72 (2010-11-09)
To evaluate whether the same or different patients respond to triptans and telcagepant. Telcagepant is an oral calcitonin gene-related peptide receptor antagonist with acute antimigraine efficacy comparable to oral triptans. It is currently unknown whether migraine patients who cannot be
Serotonergic stimulation induces nerve growth and promotes visual learning via posterior eye grafts in a vertebrate model of induced sensory plasticity
Blackiston DJ, et al.
NPJ Regenerative medicine, 2(1), 8-8 (2017)
B Lee Peterlin et al.
Expert opinion on drug metabolism & toxicology, 3(6), 899-911 (2007-11-22)
Migraine is a common, often disabling, neurovascular disease that has been shown to be associated with abnormal serotonergic activity. Drugs that modulate serotonin receptors are commonly used in the acute treatment of a migraine attack. Zolmitriptan, a 5-hydroxytryptophan(1B/1D) receptor agonist

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service