Skip to Content
Merck
All Photos(1)

Documents

L0595000

Lidocaine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Diethylamino-N-(2,6-dimethylphenyl)acetamide, Lignocaine, Xylocaine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22N2O
CAS Number:
Molecular Weight:
234.34
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

lidocaine

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCN(CC)CC(=O)Nc1c(C)cccc1C

InChI

1S/C14H22N2O/c1-5-16(6-2)10-13(17)15-14-11(3)8-7-9-12(14)4/h7-9H,5-6,10H2,1-4H3,(H,15,17)

InChI key

NNJVILVZKWQKPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lidocaine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Na+ channel blocker; class IB antiarrhythmic that is rapidly absorbed after parenteral administration.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbara Steblaj et al.
Veterinary anaesthesia and analgesia, 41(6), 583-591 (2014-08-08)
To compare pulmonary function and gas exchange in ponies during maintenance of anaesthesia with isoflurane or by a total intravenous anaesthesia (TIVA) technique. Experimental, cross-over study. Six healthy ponies weighing mean 286 (range 233-388) ± SD 61 kg, age 13
Faith A Bazley et al.
IEEE transactions on neural systems and rehabilitation engineering : a publication of the IEEE Engineering in Medicine and Biology Society, 22(5), 953-964 (2014-05-08)
The adult central nervous system is capable of significant reorganization and adaptation following neurotrauma. After a thoracic contusive spinal cord injury (SCI) neuropathways that innervate the cord below the epicenter of injury are damaged, with minimal prospects for functional recovery.
Bartholomeus C M Benno Haarman et al.
Brain, behavior, and immunity, 40, 219-225 (2014-04-08)
The "monocyte-T-cell theory of mood disorders" regards neuroinflammation, i.e. marked activation of microglia, as a driving force in bipolar disorder. Microglia activation can be visualized in vivo using [(11)C]-(R)-PK11195 PET. Indirect evidence suggests the hippocampus as a potential focus of
Song-Young Park et al.
American journal of physiology. Heart and circulatory physiology, 307(3), H346-H352 (2014-06-08)
Unlike cardiac and skeletal muscle, little is known about vascular smooth muscle mitochondrial respiration. Therefore, the present study examined mitochondrial respiratory rates in smooth muscle of healthy human feed arteries and compared with that of healthy cardiac and skeletal muscles.
Jason Wasiak et al.
The Cochrane database of systematic reviews, 6(6), CD005622-CD005622 (2012-06-15)
This is an update of the review on 'Lidocaine for pain relief in burn injured patients' first published in Issue 3, 2007. Pain is a major issue for patients suffering from many different types of wounds, in particular those with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service