Skip to Content
Merck
All Photos(1)

Key Documents

K4394

Sigma-Aldrich

PKF118-310

≥98% (HPLC)

Synonym(s):

1,6-Dimethyl-pyrimido[5,4-e]-1,2,4-triazine-5,7(1H,6H)-dione, Toxoflavin, Xanthothricin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7N5O2
CAS Number:
Molecular Weight:
193.16
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

H2O: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN1N=CN=C(C1=N2)C(N(C)C2=O)=O

InChI

1S/C7H7N5O2/c1-11-6(13)4-5(10-7(11)14)12(2)9-3-8-4/h3H,1-2H3

InChI key

SLGRAIAQIAUZAQ-UHFFFAOYSA-N

Biochem/physiol Actions

PKF118-310 is an antagonist of the Tcf4/b-catenin signaling. The compound disrupts the Tcf4/b-catenin complex and inhibits expression of Tcf4 responsive genes. PKF118-310 inhibits expression of survivin and induces apoptosis in HCC, colon tumor and lymphocytic leukemia cell lines.
PKF118-310 is a Tcf4/b-catenin antagonist.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boknam Jung et al.
Nature communications, 9(1), 31-31 (2018-01-04)
Bacterial-fungal interactions are widely found in distinct environments and contribute to ecosystem processes. Previous studies of these interactions have mostly been performed in soil, and only limited studies of aerial plant tissues have been conducted. Here we show that a
Ji-Eun Choi et al.
PloS one, 13(1), e0183893-e0183893 (2018-01-03)
Toxoflavin, a 7-azapteridine phytotoxin produced by the bacterial pathogens such as Burkholderia glumae and Burkholderia gladioli, has been known as one of the key virulence factors in crop diseases. Because the toxoflavin had an antibacterial activity, a metagenomic E. coli
T Nagamatsu et al.
Chemical & pharmaceutical bulletin, 41(2), 362-368 (1993-02-01)
6-Phenyl analogs of toxoflavin (1-methyl-6-phenylpyrimido[5,4-e]-1,2,4-triazine-5,7(1H,6H)-diones ) (7a--f) and their 4-oxides (8a-f) were synthesized by nitrosative or nitrative cyclization of the aldehyde hydrazones (6a-f) of 6-(1-methylhydrazino)-3-phenyluracil (5). Both sets of compounds, 7a-f and 8a-f, gave the corresponding 1-demethyl derivatives (10a-f) upon
Jinwoo Kim et al.
Molecular microbiology, 54(4), 921-934 (2004-11-04)
Burkholderia glumae BGR1 produces a broad-host range phytotoxin, called toxoflavin, which is a key pathogenicity factor in rice grain rot and wilt in many field crops. Our molecular and genetic analyses of toxoflavin-deficient mutants demonstrated that gene clusters for toxoflavin
Phyllis S Y Chong et al.
Oncogene, 38(9), 1508-1519 (2018-10-12)
Aberrant activation of Wnt/β-catenin signaling pathway is essential for the development of AML; however, the mechanistic basis for this dysregulation is unclear. PRL-3 is an oncogenic phosphatase implicated in the development of LSCs. Here, we identified Leo1 as a direct

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service