Skip to Content
Merck
All Photos(1)

Key Documents

K3750

Sigma-Aldrich

L-Kynurenine sulfate salt

≥98% (HPLC), suitable for ligand binding assays

Synonym(s):

β-Anthraniloyl-L-alanine, L-2-Amino-4-(2-aminophenyl)-4-oxobutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H12N2O3 · H2SO4
CAS Number:
Molecular Weight:
306.29
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

L-Kynurenine sulfate salt, crystalline

Assay

≥98% (HPLC)

Quality Level

form

crystalline

technique(s)

ligand binding assay: suitable

color

light yellow

storage temp.

−20°C

SMILES string

OS(O)(=O)=O.N[C@@H](CC(=O)c1ccccc1N)C(O)=O

InChI

1S/C10H12N2O3.H2O4S/c11-7-4-2-1-3-6(7)9(13)5-8(12)10(14)15;1-5(2,3)4/h1-4,8H,5,11-12H2,(H,14,15);(H2,1,2,3,4)/t8-;/m0./s1

InChI key

KAXRWMOLNJZCEW-QRPNPIFTSA-N

Application

L-Kynurenine sulfate salt has been used as a substrate to study the enzyme activity of kynurenine aminotransferase. It has also been used in the synthesis of Kyn adducts of certain amino acids .

Biochem/physiol Actions

Key intermediate in the breakdown pathway of tryptophan.
L-Kynurenine (L-Kyn) is a precursor of kynurenic acid which is the only recognized endogenous excitatory amino acid receptor antagonist in the central nervous system. L-Kyn is known to be a pigment generating component in animals. In mammals, it modulates the transmission of glutamate neurotransmitter. It is also considered to be a sex-attracting pheromone in vertebrates.
L-Kynurenine is a key intermediate in the breakdown of L-tryptophan and the formation of nicotinamide adenine dinucleotide (NAD+) via the kynurenine pathway. L-kynurenine is involved in a variety of neurological processes and diseases. L-kynurenine is a substrate for kynureninase/kynurenine hydrolase; kynurenine 3-monooxygenase and kynurenine-oxoglutarate transaminase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

L-Kynurenine, an amino acid identified as a sex pheromone in the urine of ovulated female masu salmon.
Yambe H
Proceedings of the National Academy of Sciences of the USA, 103(42), 15370-15374 (2006)
K Nagy et al.
Bioorganic & medicinal chemistry, 19(24), 7590-7596 (2011-11-15)
The overactivation of excitatory amino acid receptors plays a key role in the pathomechanism of several neurodegenerative disorders and in ischemic and post-ischemic events. Kynurenic acid (KYNA) is an endogenous product of the tryptophan metabolism and, as a broad-spectrum antagonist
Neuroprotective effects of L-kynurenine on hypoxia-ischemia and NMDA lesions in neonatal rats.
Nozaki K and Beal MF
Journal of Cerebral Blood Flow and Metabolism, 12(3), 400-407 (1992)
Mitochondrial aspartate aminotransferase: a third kynurenate-producing enzyme in the mammalian brain.
Guidetti P
Journal of Neurochemistry, 102(1), 103-111 (2007)
Klas Linderholm et al.
International journal of tryptophan research : IJTR, 3, 1-12 (2010-01-01)
Kynurenic acid (KYNA) is an endogenous metabolite of tryptophan. Studies have revealed increased brain KYNA levels in patients with schizophrenia. Prepulse inhibition (PPI) is a behavioral model for sensorimotor gating and found to be reduced in schizophrenia. Previous studies have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service