Skip to Content
Merck
All Photos(2)

Key Documents

K3394

Sigma-Aldrich

KH7

≥98% (HPLC)

Synonym(s):

(E)-2-(1H-Benzo[d]imidazol-2-ylthio)-N′-(5-bromo-2-hydroxybenzylidene)propanehydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15BrN4O2S
CAS Number:
Molecular Weight:
419.30
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

CC(Sc1nc2ccccc2[nH]1)C(=O)N\N=C\c3cc(Br)ccc3O

InChI

1S/C17H15BrN4O2S/c1-10(25-17-20-13-4-2-3-5-14(13)21-17)16(24)22-19-9-11-8-12(18)6-7-15(11)23/h2-10,23H,1H3,(H,20,21)(H,22,24)/b19-9+

InChI key

WILMXUAKQKGGCC-DJKKODMXSA-N

Application

KH7 has been used:
  • as a soluble adenyl cyclase (sAC) antagonist to study its effect on the signaling pathway mediated by the action of transient  receptor type 1 (TRPV1) in sperm cells.
  • as a selective sAC antagonist to study its effect on the signaling pathway of proton gated channels (HV1) induced action in sperm cells.
  • as a sAC inhibitor to study its effects on cAMP increase in monophosphorylated myosin light chain (pMLC).

Biochem/physiol Actions

KH7 is a selective inhibitor of soluble adenylyl cyclase. Soluble adenylyl cyclase (sAC) is an ubiquitously expressed, essential component of cAMP-signaling.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Adenylyl cyclases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Suitability

KH7 is not active against sAC in the presence of detergents. When used in cellular experiments, KH7 exhibits non-specific membrane disruption effects at concentrations above 50 μM.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nanshan Song et al.
Biochemical pharmacology, 150, 245-255 (2018-02-16)
β2-Aderenergic receptor (β2AR) agonist, Salmeterol exhibits anti-inflammatory activities. However, the inhibitory effects of Salmeterol on inflammasome activation are elusive and the underlying mechanisms need to be explored. In this study, we established inflammatory model in primary bone marrow-derived macrophages (BMDM)
Carresse L Gerald et al.
Alcoholism, clinical and experimental research, 40(2), 273-283 (2016-02-05)
Farm workers in rural areas consume more alcohol than those who reside in urban areas. Occupational exposures such as agricultural work can pose hazards on the respiratory system. It is established that hog barn dust induces inflammation in the airway
Frances Evans et al.
Journal of cellular physiology, 235(3), 2947-2962 (2019-09-20)
In previous work, we reported that plasma membrane potential depolarization (PMPD) provokes cortical F-actin remodeling in bovine corneal endothelial (BCE) cells in culture, which eventually leads to the appearance of intercellular gaps. In kidney epithelial cells it has been shown
Mandy Lo et al.
Journal of comparative physiology. B, Biochemical, systemic, and environmental physiology, 191(1), 113-125 (2020-11-21)
We tested in six fish species [Pacific lamprey (Lampetra richardsoni), Pacific spiny dogfish (Squalus suckleyi), Asian swamp eel (Monopterus albus), white sturgeon (Acipenser transmontanus), zebrafish (Danio rerio), and starry flounder (Platichthys stellatus)] the hypothesis that elevated extracellular [HCO3-] protects spontaneous
Xiudan Wang et al.
Fish & shellfish immunology, 66, 140-147 (2017-05-10)
Ocean acidification (OA) has deleterious impacts on immune response and energy homeostasis status of Mollusca. In the present study, the apoptosis ratio of hemocytes and the adenosine triphosphate (ATP) allocation in gill tissues were determined after Pacific oysters Crassostrea gigas

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service