Skip to Content
Merck
All Photos(1)

Key Documents

S153

Sigma-Aldrich

SQ 22,536

≥97% (HPLC), powder

Synonym(s):

9-(Tetrahydro-2-furanyl)-9H-purin-6-amine, 9-THF-Ade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11N5O
CAS Number:
Molecular Weight:
205.22
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >10 mg/mL
H2O: insoluble

SMILES string

Nc1ncnc2n(cnc12)C3CCCO3

InChI

1S/C9H11N5O/c10-8-7-9(12-4-11-8)14(5-13-7)6-2-1-3-15-6/h4-6H,1-3H2,(H2,10,11,12)

InChI key

UKHMZCMKHPHFOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

SQ 22,536 was used to study the role of adenylate cyclase in differentiation of PC12 cells and in gap junctional intercellular communication in breast cancer cells.

Biochem/physiol Actions

SQ 22,536 is an effective inhibitor of not only basal but also prosptaglandin E1-activated adenylate cyclase activities in platelets.1 It reverses hyperalgesia contralaterally and ipsilaterally when injected intramuscularly in rats.2
SQ 22,536 is a cell-permeable adenylyl cyclase inhibitor. IC50 = 20 μM in detergent-dispersed rat brain preparation.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Adenylyl cyclases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Haslam et al.
The Biochemical journal, 176(1), 83-95 (1978-10-15)
Whereas adenosine itself exerted independent stimulatory and inhibitory effects on the adenylate cyclase activity of a platelet particulate fraction at low and high concentrations respectively, 2-substituted and N6-monosubstituted adenosines had stimulatory but greatly decreased inhibitory effects. Deoxyadenosines, on the other
Marie K Hoeger-Bement et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 23(13), 5437-5445 (2003-07-05)
Spinal activation of the cAMP pathway produces mechanical hyperalgesia, sensitizes nociceptive spinal neurons, and phosphorylates the transcription factor cAMP-responsive element binding protein (CREB), which initiates gene transcription. This study examined the role of the cAMP pathway in a model of
Hari Hendarto et al.
Metabolism: clinical and experimental, 61(10), 1422-1434 (2012-05-05)
Accumulating evidence has implicated that GLP-1 may have a beneficial effect on cardiovascular and renal diseases but the mechanism is not fully understood. Here we show that GLP-1 analog, liraglutide, inhibits oxidative stress and albuminuria in streptozotocin (STZ)-induced type 1
Soo Hyeon Kim et al.
Blood, 120(24), 4892-4902 (2012-10-18)
In cancer, VEGF-induced increase in vascular permeability results in increased interstitial pressure, reducing perfusion and increasing hypoxia, which reduce delivery of chemotherapeutic agents and increase resistance to ionizing radiation. Here, we show that both TIMP-2 and Ala + TIMP-2, a
Sangwoo Shim et al.
Proceedings of the National Academy of Sciences of the United States of America, 120(23), e2220037120-e2220037120 (2023-05-30)
The balance between neural stem cell proliferation and neuronal differentiation is paramount for the appropriate development of the nervous system. Sonic hedgehog (Shh) is known to sequentially promote cell proliferation and specification of neuronal phenotypes, but the signaling mechanisms responsible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service