Skip to Content
Merck
All Photos(3)

Documents

862169

Sigma-Aldrich

2-cis,4-trans-Abscisic acid

synthetic, 98%

Synonym(s):

(±)-Abscisic acid, (2Z,4E)-5-(1-Hydroxy-2,6,6-trimethyl-4-oxo-2-cyclohexen-1-yl)-3-methyl-2,4-pentadienoic acid, ABA, Dormin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O4
CAS Number:
Molecular Weight:
264.32
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

186-188 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(\C=C\C1(O)C(C)=CC(=O)CC1(C)C)=C\C(O)=O

InChI

1S/C15H20O4/c1-10(7-13(17)18)5-6-15(19)11(2)8-12(16)9-14(15,3)4/h5-8,19H,9H2,1-4H3,(H,17,18)/b6-5+,10-7-

InChI key

JLIDBLDQVAYHNE-LXGGSRJLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-cis,4-trans-Abscisic acid (ABA) is an organic compound used as a starting material in in organic synthesis. It is also used as a chiral auxiliary during asymmetric synthesis. ABA derivatives can be used to control the stereochemistry of reactions by serving as chiral templates that induce the formation of specific products.

Application


  • Chiral Separation of Abscisic Acid: Research demonstrated direct chiral separation of abscisic acid using high-performance liquid chromatography with a phenyl column and a gamma-cyclodextrin mobile phase, advancing the analytical methods essential for studying plant stress hormones (Terashima et al., 2023).

  • Xanthine Oxidase Inhibitory Activity: A study including abscisic acid derivatives showed significant xanthine oxidase inhibitory activity, offering potential health benefits and therapeutic applications (Miyata et al., 2019).

  • Anticancer Potential: Extracts containing abscisic acid from mangrove endophytic fungi exhibited cytotoxic properties and induced G(0)/G(1) cell cycle arrest and apoptosis in human cancer cells, highlighting their potential in cancer treatment strategies (Zhou et al., 2018).

  • Antidiabetic Activity: Extracts involving abscisic acid from an endophytic fungus showed promising antidiabetic activity, potentially improving treatment options for diabetes through natural product research (Uzor et al., 2017).

  • Glucose Tolerance and Insulinemia Reduction: A groundbreaking study found that microgram amounts of abscisic acid in fruit extracts significantly improve glucose tolerance and reduce insulin levels in rats and humans, suggesting its use as a dietary supplement for managing diabetes (Magnone et al., 2015).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott Hayes et al.
Current biology : CB, 29(10), 1669-1676 (2019-05-06)
Global food production is set to keep increasing despite a predicted decrease in total arable land [1]. To achieve higher production, denser planting will be required on increasingly degraded soils. When grown in dense stands, crops elongate and raise their
Mirko Magnone et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(12), 4783-4793 (2015-08-06)
2-Cis,4-trans-abscisic acid (ABA) is a plant hormone that is present also in animals. Several lines of evidence suggest that ABA contributes to the regulation of glycemia in mammals: nanomolar ABA stimulates insulin release from β-pancreatic cells and glucose transporter-4-mediated glucose
Haifeng Jia et al.
The New phytologist, 198(2), 453-465 (2013-02-22)
Fleshy fruits are classically divided into climacteric and nonclimacteric types. It has long been thought that the ripening of climacteric and nonclimacteric fruits is regulated by ethylene and abscisic acid (ABA), respectively. Here, we report that sucrose functions as a
Kamil Frankowski et al.
Postepy biochemii, 59(1), 83-88 (2013-07-05)
Abscisic acid is one of the plant hormones that determines normal growth and development, i.e. seeds ripening and germination, stomata opening and closure, flowering and stress responses. An appropriate level of endogenous ABA plays a key role in the regulation
Jean-Luc Montillet et al.
PLoS biology, 11(3), e1001513-e1001513 (2013-03-26)
Plant stomata function in innate immunity against bacterial invasion and abscisic acid (ABA) has been suggested to regulate this process. Using genetic, biochemical, and pharmacological approaches, we demonstrate that (i) the Arabidopsis thaliana nine-specific-lipoxygenase encoding gene, LOX1, which is expressed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service