Skip to Content
Merck
All Photos(1)

Key Documents

I3391

Sigma-Aldrich

Anti-Human IgG (Fc specific), F(ab′)2 fragment antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat anti-human IgG

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

concentration

2.0 mg/mL protein

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

IgG antibody plays a crucial role in humoral immune responses such as phagocytosis, complement activation, placental transport and cell surface-receptor binding. Anti-human IgG (Fc specific), F(ab′)2 fragment antibody can be used as a capture antibody in antibody-binding assay for soluble ICAM-1-Fc. Goat anti-human-IgG (Fc specific) F(ab′)2 antibody reacts specifically with human IgG but does not react with other immunoglobulins.
IgG is the main type of antibody in human serum, secreted by the B cells. It consists of four polypeptide chains, made of two identical 50 kDa γ heavy (H) chains and two identical 25 kDa κ or λ light (L) chains, which are linked by inter-chain disulfide bonds. IgG contains four isotypes IgG1, IgG2, IgG3 and IgG4.

Immunogen

Purified human IgG

Application

Anti-Human IgG (Fc specific), F(ab′)2 fragment antibody produced in goat has been used in studying the antibody binding assay of soluble intercellular adhesion molecule-1 (ICAM-1)-Fc.
Anti-human IgG (Fc specific), F(ab′)2 fragment antibody can be used in ELISA to determine human IgG1-Fc protein.

Biochem/physiol Actions

IgG stimulates the classical pathway of the complement system. It neutralizes virus particles and toxins. IgG plays a crucial role in antibody-dependent cell-mediated cytotoxicity (ADCC). IgG might be associated with allergy. It has the longest serum half life.

Other Notes

Antibody adsorbed with mouse and rat IgG

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 0.05% sodium azide

Preparation Note

Adsorbed to reduce background with mouse or rat samples.
Useful when trying to avoid background staining due to the presence of Fc receptors.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gang Song et al.
The Journal of biological chemistry, 281(8), 5042-5049 (2005-12-16)
The interaction between integrin lymphocyte function-associated antigen-1 (LFA-1) and its ligand intercellular adhesion molecule-1 (ICAM-1) is critical in immunological and inflammatory reactions but, like other adhesive interactions, is of low affinity. Here, multiple rational design methods were used to engineer
Yusuke Higuchi et al.
Nature communications, 12(1), 3802-3802 (2021-06-23)
SARS-CoV-2 has mutated during the global pandemic leading to viral adaptation to medications and vaccinations. Here we describe an engineered human virus receptor, ACE2, by mutagenesis and screening for binding to the receptor binding domain (RBD). Three cycles of random
Rational design of intercellular adhesion molecule-1 (ICAM-1) variants for antagonizing integrin lymphocyte function-associated antigen-1-dependent adhesion
Song G, et al.
The Journal of Biological Chemistry, 281(8), 5042-5049 (2006)
Leander Blaas et al.
BMC biotechnology, 9, 3-3 (2009-01-16)
The development of appropriate expression vectors for large scale protein production constitutes a critical step in recombinant protein production. The use of conventional expression vectors to obtain cell lines is a cumbersome procedure. Often, stable cell lines produce low protein
Camila A Wilkens et al.
PloS one, 10(3), e0119053-e0119053 (2015-03-15)
Cell engineering has been used to improve animal cells' central carbon metabolism. Due to the central carbon metabolism's inefficiency and limiting input of carbons into the TCA cycle, key reactions belonging to these pathways have been targeted to improve cultures'

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service