Skip to Content
Merck
All Photos(1)

Key Documents

I5260

Sigma-Aldrich

Anti-Human IgG (Fab specific) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Fab-Specific Antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

indirect ELISA: 1:50,000
quantitative precipitin assay: 2.0 mg/mL

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Anti-Human IgG (Fab specific) antiserum is produced in goat using purified human IgG Fab fragment as the immunogen. Affinity isolated antibody is obtained from goat anti-human IgG antiserum by immunospecific purification which removes essentially all goat serum proteins, including immunoglobulins, which do not specifically bind to the Fab fragment of human IgG. IgG is a larger molecule with a molecular weight of 150 kilo daltons (kDa). IgG has two light chains (L) and two heavy chains (H). The light chain can be either lambda (λ) or kappa (κ) chain. Functionally, immunoglobulins have constant region and variable region. Light chain has one variable region in the N-terminal VL and followed by one constant domain CL. The light chain is approximately about 25 kDa. The heavy chain has one variable region in the N-terminal and followed by three constant domains. In between the first (C1H) and second constant (C2H) domain in the spacer hinge region which connects both the heavy chains. Each heavy chain is about 55 kDa.

Specificity

Goat anti-human IgG (Fab specific) antibody reacts specifically with Fab fragment of human IgG and shows no reactivity for Fc fragment of human IgG. This product has no interspecies cross reactivity for mouse and rat IgG.

Immunogen

Purified human IgG Fab fragment.

Application

Anti-Human IgG (Fab specific) antibody produced in goat is effective as a second antibody reagent in immunoassay procedures. It may be used:
  • as starting material for conjugates using enzymes or fluorescent dyes
  • in enzyme linked immunosorbent assay
  • as a cross-linking antibody in internalisation of 125I-IgG by streptolysin-O-permeabilised cells
  • as capture antibody in enzyme-linked immunosorbent assay (ELISA) for detection and semiquantitation of Fab
  • in electrophoresis and immunoelectrophoresis

Biochem/physiol Actions

Enzymatic cleavage of IgG yields antigen-binding (Fab) and effector activating (Fc) fragments. IgG treatment with papain produces two Fab fragments and one Fc fragment. IgG treatment with pepsin produces (Fab′)2 fragment alone. IgG antibodies regulate several functions such as complement activation and phagocytosis. Thus they play a crucial role in facilitating cytological immune responses. Anti-human IgG (Fab specific) antibody can be used in Ouchterlony double diffusion. Due to lack of interspecies cross reactivity to mouse or rat ascites fluids, this antibody is also ideal for screening human monoclonal antibodies produced by hybridoma cells grown in vivo.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide as preservative

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard J Kraus et al.
Journal of virology, 94(18) (2020-07-10)
We previously reported that the cellular transcription factor hypoxia-inducible factor 1α (HIF-1α) binds a hypoxia response element (HRE) located within the promoter of Epstein-Barr virus's (EBV's) latent-lytic switch BZLF1 gene, Zp, inducing viral reactivation. In this study, EBV-infected cell lines
Human recombinant antibodies against Plasmodium falciparum merozoite surface protein 3 cloned from peripheral blood leukocytes of individuals with immunity to malaria demonstrate antiparasitic properties
Lundquist R, et al.
Infection and Immunity, 74(6), 3222-3231 (2006)
Edurne Rujas et al.
Cell reports, 32(7), 108037-108037 (2020-08-20)
The contribution of membrane interfacial interactions to recognition of membrane-embedded antigens by antibodies is currently unclear. This report demonstrates the optimization of this type of antibodies via chemical modification of regions near the membrane but not directly involved in the
Endocytosis of FcgammaRI is regulated by two distinct signalling pathways
Norman JC and Allen JM
Febs Letters, 484(3), 179-183 (2000)
Qi Yang et al.
Analytical chemistry, 88(23), 11804-11812 (2016-11-03)
Circulating antibodies (Ab) that specifically bind polyethylene glycol (PEG), a biocompatible polymer routinely used in protein and nanoparticle therapeutics, have been associated with reduced efficacy of and/or adverse reactions to therapeutics modified with or containing PEG. Unlike most antidrug antibodies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service