P8651
Protein A–Peroxidase from Staphylococcus aureus/horseradish
lyophilized powder
Synonym(s):
Peroxidase–Protein A
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
conjugate
peroxidase conjugate
Quality Level
100
200
form
lyophilized powder
technique(s)
direct ELISA: 1:80,000 using human IgG
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Protein A is a cell wall constituent of Staphylococcus aureus, with a molecular weight of 56kDa. It possesses five binding domains which interact with heavy chains of immunoglobulins at the Fc or Fab fragment. In response to protein A, the complement system, interferon production, hypersensitivity reactions and cell-mediated cytotoxicity get activated. Horseradish peroxidase is a heme-containing enzyme. In the presence of hydrogen peroxide, the enzyme catalyzes the oxidation of luminol.
Specificity
Binds IgG only from most mammals, except rat, goat, sheep.
Application
Protein A–Peroxidase from Staphylococcus aureus/horseradish has been used in enzyme-linked immunosorbent assay (ELISA).
Packaging
Package size based on protein content.
Physical form
Lyophilized powder containing sodium citrate buffer salt. Protein determined by A205.
Preparation Note
Protein A, extracellular, coupled to Peroxidase, Type VI, by a modification of the procedure of O′Sullivan, M.J., et al., FEBS Lett., 95, 311 (1978), which favors low molecular weight conjugates.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Biosensors for Direct Monitoring of Environmental Pollutants in Field (2013)
Bioconjugate Techniques (2013)
Journal of immunological methods, 218(1-2), 123-132 (1998-11-18)
We have developed a dot immunobinding assay (CM) for the serodiagnosis of the human hydatidosis. The assay employs Samaron blue colloidal dye particles conjugated to hydatid antigen (HA) or protein A, both of which serve as visualizing agents. The test
The Plant cell, 21(10), 3170-3184 (2009-10-27)
The plant hormone abscisic acid (ABA) orchestrates plant adaptive responses to a variety of stresses, including drought. This signaling pathway is regulated by reversible protein phosphorylation, and genetic evidence demonstrated that several related protein phosphatases 2C (PP2Cs) are negative regulators
Identification of a Complex of the Three Forms of the Rat Liver
Asialoglycoprotein Receptor
Asialoglycoprotein Receptor
The Journal of Biological Chemistry, 263 (1998)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service