Skip to Content
Merck
All Photos(3)

Key Documents

I0899

Sigma-Aldrich

Imipramine hydrochloride

BioXtra, ≥99% (TLC)

Synonym(s):

10,11-Dihydro-N,N-dimethyl-5H-dibenz[b,f]azepine-5-propanamine hydrochloride, 5-[3-(Dimethylamino)propyl]-10,11-dihydro-5H-dibenz[b,f]azepine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24N2 · HCl
CAS Number:
Molecular Weight:
316.87
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.25

product line

BioXtra

Assay

≥99% (TLC)

form

powder

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

solubility

H2O: 50 mg/mL, colorless to faintly yellow

anion traces

sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

Cl[H].CN(C)CCCN1c2ccccc2CCc3ccccc13

InChI

1S/C19H24N2.ClH/c1-20(2)14-7-15-21-18-10-5-3-8-16(18)12-13-17-9-4-6-11-19(17)21;/h3-6,8-11H,7,12-15H2,1-2H3;1H

InChI key

XZZXIYZZBJDEEP-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application


  • Novel, fast, and reliable electrochemical dsDNA biosensor based on O-terminated pristine nanocrystalline boron-doped diamond electrode for DNA interaction studies.: Research featuring a novel electrochemical dsDNA biosensor that facilitates the study of DNA interactions with imipramine hydrochloride, enhancing our understanding of its genotoxic effects and providing crucial insights for biochemistry applications (Augustín M et al., 2024).

  • Comparison of two pharmacological protocols for inducing ex copula ejaculation in donkeys.: In this comparative study, imipramine hydrochloride is used within a pharmacological protocol, offering significant insights into veterinary applications and animal reproductive studies (Khan FA et al., 2024).

  • Development of Visible Spectrophotometric Methods for the Determination of Tricyclic Antidepressants Based on Formation of Molecular Complexes with p-Benzoquinones.: This publication outlines a novel spectrophotometric method for analyzing tricyclic antidepressants like imipramine hydrochloride, providing an efficient tool for quantitative analysis in pharmaceutical and clinical research (Ciuca MD et al., 2023).

Biochem/physiol Actions

Tricyclic antidepressant; inhibits the serotonin and norepinephrine transporters with Kis of 7.7 nM and 67 nM, respectively. Has little effect on the dopamine transporter (Ki = 25 μM).
Tricyclic antidepressant; inhibits the serotonin and norepinephrine transporters; has little effect on the dopamine transporter.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steven Chen et al.
Communications biology, 3(1), 747-747 (2020-12-23)
Approximately 10% of the world's population is at risk of schistosomiasis, a disease of poverty caused by the Schistosoma parasite. To facilitate drug discovery for this complex flatworm, we developed an automated high-content screen to quantify the multidimensional responses of
Adam J Walker et al.
Behavioural brain research, 242, 76-83 (2013-01-02)
Several animal models are currently utilised in the investigation of major depressive disorder; however, each is validated by its response to antidepressant pharmacotherapy. Few animal models consider the notion of antidepressant treatment resistance. Chronic daily administration of adrenocorticotropic hormone (ACTH)
Ana Laura de la Garza et al.
Nutrients, 11(3) (2019-03-15)
Maternal hypercaloric exposure during pregnancy and lactation is a risk factor for developing diseases associated with inflammation such as obesity, diabetes and, neurological diseases in the offspring. Neuroinflammation might modulate neuronal activation and flavonoids are dietary compounds that have been
Alexander Carpinteiro et al.
Cell reports. Medicine, 1(8), 100142-100142 (2020-11-10)
The acid sphingomyelinase/ceramide system plays an important role in bacterial and viral infections. Here, we report that either pharmacological inhibition of acid sphingomyelinase with amitriptyline, imipramine, fluoxetine, sertraline, escitalopram, or maprotiline or genetic downregulation of the enzyme prevents infection of
Management of nocturnal enuresis.
Patrina H Y Caldwell et al.
BMJ (Clinical research ed.), 347, f6259-f6259 (2013-10-31)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service