Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
usage
sufficient for 100 reactions
feature
dNTPs included, hotstart
technique(s)
RT-PCR: suitable using : suitable, cDNA synthesis: suitable using : suitable, WGA
color
colorless
input
purified RNA
shipped in
wet ice
storage temp.
−20°C
General description
One-step: In a single tube, eAMV™ RT and AccuTaq LA act sequentially to first produce cDNA and then immediately amplify by PCR. This provides quick, sensitive analysis of RNA.
Two-step: Each reaction is individually optimized for greater yields with high fidelity, when protocol requires multiple amplifications, or if maximum yield is more important than maximum convenience.
Application
Features and Benefits
- Greater transcription lengths than other reverse transcriptases, generating cDNA up to 14.1 Kb.
- Higher sensitivity for detecting low abundance messages. eAMV™ RT is able to transcribe RNA that other reverse transcriptases cannot detect.
- Unsurpassed transcription through difficult secondary structure.
- Increased sensitivity, specificity and yield from JumpStart AccuTaq LA DNA Polymerase.
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description Flexible kit for one-step or two-step RT-PCR | description Components for cDNA synthesis with enhanced AMV reverse transcriptase | description One step SYBR™ Green RT-qPCR with MMLV & hot-start Taq DNA Polymerase | description sufficient for 50 reactions, sufficient for 150 reactions, suitable for RT-PCR, hotstart |
| technique(s) RT-PCR: suitable, cDNA synthesis: suitable | technique(s) RT-PCR: suitable, cDNA synthesis: suitable | technique(s) RT-qPCR: suitable | technique(s) RT-PCR: suitable, RT-qPCR: suitable |
| usage sufficient for 100 reactions | usage sufficient for 50 reactions, kit sufficient for 50 reactions | usage sufficient for 250 reactions | usage sufficient for 150 reactions, sufficient for 50 reactions |
| shipped in wet ice | shipped in dry ice | shipped in wet ice | shipped in - |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level - |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. - |
| feature dNTPs included, hotstart | feature dNTPs included, hotstart: no | feature dNTPs included, hotstart | feature hotstart |
Storage Class
10 - Combustible liquids
flash_point_f
No data available
flash_point_c
No data available
signalword
Danger
wgk
WGK 3
hcodes
Hazard Classifications
Aquatic Chronic 3 - ED ENV 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



