Skip to Content
Merck
All Photos(6)

Key Documents

HPA041275

Sigma-Aldrich

Anti-SYNCRIP antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GRY-RBP, Anti-HNRNPQ, Anti-HNRPQ1, Anti-dJ3J17.2, Anti-hnRNP-Q

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

GNGTEEPMDTTSAVIHSENFQTLLDAG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SYNCRIP(10492)

General description

Synaptotagmin-binding cytoplasmic RNA-interacting protein (SYNCRIP) belongs to the heterogeneous nuclear ribonucleoprotein (hnRNP) family. This protein is located in the cytoplasm and is made up of three conserved RNA recognition motif (RRM) domains, an N-terminal domain, and a long, unstructured C-terminal domain. The SYNCRIP gene is located on the human chromosome at 6q14.3.

Immunogen

synaptotagmin binding, cytoplasmic RNA interacting protein

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-SYNCRIP antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

Synaptotagmin-binding cytoplasmic RNA-interacting protein (SYNCRIP) functions as an RNA-binding protein and is involved in muscular and neuronal development in mammals and Drosophila. It plays a role in the RNA replication of mouse hepatitis virus (MHV) and hepatitis C virus (HCV). Dysregulation of the SYNCRIP gene leads to neurodegenerative disorders and severe cardiomyopathies.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST82407

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helene Minyi Liu et al.
Virology, 386(2), 249-256 (2009-02-24)
Hepatitis C virus (HCV) RNA replication requires viral nonstructural proteins as well as cellular factors. Recently, a cellular protein, synaptotagmin-binding, cytoplasmic RNA-interacting protein (SYNCRIP), also known as NSAP1, was found to bind HCV RNA and enhance HCV IRES-dependent translation. We
Keum S Choi et al.
Journal of virology, 78(23), 13153-13162 (2004-11-16)
Several cellular proteins, including several heterogeneous nuclear ribonucleoproteins (hnRNPs), have been shown to function as regulatory factors for mouse hepatitis virus (MHV) RNA synthesis as a result of their binding to the 5' and 3' untranslated regions (UTRs) of the
Miriam Pacetti et al.
Disease models & mechanisms, 15(4) (2022-03-05)
The cellular level of TDP-43 (also known as TARDBP) is tightly regulated; increases or decreases in TDP-43 have deleterious effects in cells. The predominant mechanism responsible for the regulation of the level of TDP-43 is an autoregulatory negative feedback loop.
Aafke Engwerda et al.
European journal of human genetics : EJHG, 26(10), 1478-1489 (2018-06-16)
Proximal 6q (6q11-q15) deletions are extremely rare and little is known about their phenotypic consequences. Since parents and caregivers now use social media to seek information on rare disorders, the Chromosome 6 Project has successfully collaborated with a Facebook group
Fruzsina Hobor et al.
Nature communications, 9(1), 831-831 (2018-02-28)
Exosomal miRNA transfer is a mechanism for cell-cell communication that is important in the immune response, in the functioning of the nervous system and in cancer. Syncrip/hnRNPQ is a highly conserved RNA-binding protein that mediates the exosomal partition of a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service