Skip to Content
Merck
All Photos(7)

Key Documents

HPA013859

Sigma-Aldrich

Anti-ALOX15 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-15-LOX, Anti-Arachidonate 15-lipoxygenase, Anti-Arachidonate omega-6 lipoxygenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

RTVGEDPQGLFQKHREEELEERRKLYRWGNWKDGLILNMAGAKLYDLPVDERFLEDKRVDFEVSLAKGLADLAIKDSLNVLTCWKDLDDFNRIFWCGQSKLAERVRDSWKEDALFGYQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALOX15(246)

General description

The gene ALOX15 (Arachidonate 15-lipoxygenase) is mapped to human chromosome 17p13.3.

Immunogen

Arachidonate 15-lipoxygenase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene ALOX15 encodes a non-heme iron-containing dioxygenase called as arachidonate 15-lipoxygenase. It encodes the type 1 isozyme of 15-Lipoxygenase. The enzyme catalyzes the oxygenation of free fatty acids and fatty acids bound to membrane phospholipids. The reaction converts arachidonic acid to 12- and 15-HETE (hydroxyeicosatetraenoic acid). The gene has been associated with the development of asthma, arthritis, and atherosclerosis. Polymorphism in this gene has been implicated as a determinant of bone mineral density in postmenopausal women and may serve as a marker for osteoporosis. The enzyme may play a role in inflammation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72464

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomohiko Urano et al.
Journal of bone and mineral metabolism, 23(3), 226-230 (2005-04-20)
The 12/15-lipoxygenase gene Alox15 has been identified as a susceptibility gene for bone mineral density (BMD) in mice through combined genetic and genomic analyses. Here we studied the association between bone mineral density and an ALOX15 gene single nucleotide polymorphism
Amira Gohara et al.
Oncology reports, 28(4), 1275-1282 (2012-07-25)
Lipoxygenases make an impact on every stage of cancer affecting carcinogenesis, metastasis and apoptosis. While there is a rich literature on individual lipoxygenases we lack extensive data on their coexistence and balance in different organs and types of cancer. Renal
Shoji Ichikawa et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 21(4), 556-564 (2006-04-07)
The Alox15 gene was recently identified as a negative regulator of peak BMD in mice. Polymorphisms in human ALOX12, but not ALOX15, were significantly associated with spine BMD in white men and women, suggesting that ALOX12 may contribute to normal
Jonas Wittwer et al.
Human mutation, 27(1), 78-87 (2005-12-02)
The reticulocyte-type 15-lipoxygenase-1 (ALOX15) has antiinflammatory and inflammatory effects, and is implicated in the development of asthma, arthritis, and atherosclerosis. We screened the human ALOX15 gene for variations because genetic variability in ALOX15 may influence these diseases. We detected 11
Jeffrey R Liddell et al.
Molecular neurodegeneration, 19(1), 14-14 (2024-02-06)
Ferroptosis is a form of regulated cell death characterised by lipid peroxidation as the terminal endpoint and a requirement for iron. Although it protects against cancer and infection, ferroptosis is also implicated in causing neuronal death in degenerative diseases of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service