Skip to Content
Merck
All Photos(1)

Key Documents

H7166

Sigma-Aldrich

IL-3 human

HumanKine®, recombinant, Xeno-free, >95% (SDS-PAGE), suitable for cell culture

Synonym(s):

Interleukin-3 human, IL-3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Product Name

Interleukin-3 human, IL-3, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

>95% (SDS-PAGE)

form

lyophilized powder

potency

≤2 ng/mL EC50

quality

endotoxin tested

mol wt

dimer 17-45 kDa (glycosylated)

packaging

pkg of 10 μg

manufacturer/tradename

HumanZyme

technique(s)

cell culture | mammalian: suitable

impurities

<1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL3(3562)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Interleukin-3 (IL-3) is a multifunctional protein, originally called colony forming unit-stimulating activity (CFU-SA), and is produced by activated T lymphocytes. IL-3 supports the formation of multilineage colonies in the early development of multipotent hematopoietic progenitor cells. IL-3 has been shown to induce colony formation of macrophages, neutrophils, mast cells, and megakaryocytes from agar-suspended bone marrow cells. IL-3 also interacts with IL-2 to stimulate growth of T lymphocytes and to induce IgG secretion from activated B cells. Other synonyms for IL-3 include pan-specific hemopoietin, multicolony stimulating factor, mast cell growth factor, burst promoting activity, histamine-producing cell stimulating factor, P cell stimulating factor, and WEHI-3 factor.
Interleukin-3 is a multifunctional protein produced by activated T lymphocytes. IL-3 supports the formation of multilineage colonies in the early development of multipotent hematopoietic progenitor cells. IL-3 also interacts with IL-2 to stimulate growth of T lymphocytes and induce IgG secretion from activated B cells.

Physical form

Lyophilized from a 0.2 μm filtered solution of 1x PBS.

Preparation Note

HumanKine Interleukin-3 (IL-3), expressed in human HEK 293 cells, is a glycosylated monomer with an apparent molecular mass of 17-45 kDa due to glycosylation, which is absent when this cytokine is expressed in E. coli. Production in human HEK 293 cells offers authentic glycosylation which contributes to stability in cell growth media and other applications.

Analysis Note

The activity was determined by the dosedependent stimulation of the proliferation of human TF-1 cells (human erythroleukemic indicator cell line).

Legal Information

HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S Yousefi et al.
Allergy, 70(9), 1184-1188 (2015-06-05)
Basophils are primarily associated with immunomodulatory functions in allergic diseases and parasitic infections. Recently, it has been demonstrated that both activated human and mouse basophils can form extracellular DNA traps (BETs) containing mitochondrial DNA and granule proteins. In this report
Matthew Cannon et al.
Journal of clinical medicine, 9(7) (2020-07-28)
Sickle-cell disease (SCD) is a debilitating hematological disorder with very few approved treatment options. Therapeutic reactivation of fetal hemoglobin (HbF) is one of the most pursued methods for ameliorating the systemic manifestations of SCD. Despite this, very few pharmacological agents
Amy H Lin et al.
Journal of virology, 88(17), 10066-10077 (2014-06-27)
We developed a Moloney mouse leukemia virus (MLV)-based retroviral replicating vector (RRV), Toca 511, which has displayed tumor specificity in resected brain tumor material and blood in clinical trials. Here, we investigated the interaction between Toca 511 and human host
Kerstin Kapp et al.
Molecular therapy. Nucleic acids, 3, e170-e170 (2014-06-25)
Toll-like receptors are sensing modulators of the innate immune system. One member of this protein family, Toll-like receptor (TLR)-9, is increasingly being investigated as therapeutic target for infectious diseases and cancer. Double-Stem Loop ImmunoModulator (dSLIM) is a new TLR-9 agonist
Orit Uziel et al.
Biochemical and biophysical research communications, 450(1), 274-282 (2014-06-08)
Treatment with erythropoietin (EPO) in several cancers is associated with decreased survival due to cancer progression. Due to the major importance of telomerase in cancer biology we hypothesized that some of these effects may be mediated through EPO effect on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service