Skip to Content
Merck
All Photos(2)

Documents

G5035

Sigma-Aldrich

Granulocyte-Macrophage Colony-Stimulating Factor human

≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

GM-CSF human

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

product name

Granulocyte-Macrophage Colony-Stimulating Factor human, GM-CSF, recombinant, expressed in E. coli, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

potency

≤0.1 ng/mL EC50

mol wt

14.6 kDa

packaging

pkg of 4X5 μg
pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

≤1.000 EU/μg

color

white

solubility

water: soluble 0.010 mL, clear, colorless

UniProt accession no.

storage temp.

−20°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Package size based on protein content

Biochem/physiol Actions

Granulocyte-macrophage colony stimulating factor (GM-CSF) is a growth and differentiation factor for cells in the granulocyte, macrophage and eosinophil lineage. GM-CSF stimulates colony formation from pluripotential progenitor cells at extremely low concentrations and is an essential survival and proliferative factor for hematopoietic progenitor cells in all divisions up to maturity. It also stimulates growth in some epithelial cells and osteoclasts. GM-CSF is produced by a variety of cell types (monocytes, endothelial cells, T-cells, fibroblasts, mitogen-stimulated B-cells, and LPS-stimulated macrophages). GM-CSF is secreted as a single chain glycoprotein containing 128 amino acids for human with a conserved disulfide bond. Human and murine GM-CSF share approx. 54% sequence homology and do not cross-react in bioactivity.

Physical form

Lyophilized from a 0.2 μm filtered, 10 mM sodium citrate, pH 3.5 solution.

Analysis Note

The proliferative activity of human GM-CSF is tested in culture using human TF-1 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benhur Lee et al.
Research square (2021-10-07)
Bats are significant reservoir hosts for many viruses with zoonotic potential1. SARS-CoV-2, Ebola virus, and Nipah virus are examples of such viruses that have caused deadly epidemics and pandemics when spilled over from bats into human and animal populations2,3. Careful
Alessio Lanna et al.
Methods in molecular biology (Clifton, N.J.), 2654, 251-261 (2023-04-28)
Eukaryotes solve the DNA-end replication problem synthesizing hexameric chromosome ends known as telomeres. Recent studies have uncovered unexpected functions of telomeres in linking synaptic signaling and vesicle transport, with at least one pathway directly involved in transferring telomeres through the
Satoshi Ikegame et al.
Nature microbiology, 8(6), 1108-1122 (2023-05-05)
Morbilliviruses are among the most contagious viral pathogens of mammals. Although previous metagenomic surveys have identified morbillivirus sequences in bats, full-length morbilliviruses from bats are limited. Here we characterize the myotis bat morbillivirus (MBaMV) from a bat surveillance programme in
Antonella Mancusi et al.
Blood, 125(20), 3173-3182 (2015-03-15)
Because activating killer cell immunoglobulinlike receptors (KIRs) are heterogeneously expressed in the population, we investigated the role of donor activating KIRs in haploidentical hematopoietic transplants for acute leukemia. Transplants were grouped according to presence vs absence of KIR-ligand mismatches in
Kazuhiro Maki et al.
International journal of hematology, 100(4), 361-369 (2014-07-25)
Myelodysplastic syndrome (MDS) is a stem cell tumor characterized by dysplastic features and ineffective hematopoiesis in the early phase and leukemic progression in the late phase. Speculating that differences in the expression of genes and microRNA (miRNA) in control and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service