H6764
1-Heptacosanol
≥98%
Synonym(s):
n-Heptacosanol, NSC 53829
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C27H56O
CAS Number:
Molecular Weight:
396.73
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
Assay
≥98%
storage temp.
2-8°C
SMILES string
CCCCCCCCCCCCCCCCCCCCCCCCCCCO
InChI
1S/C27H56O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27-28/h28H,2-27H2,1H3
InChI key
ULCZGKYHRYJXAU-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Sol Ji Choi et al.
Food chemistry, 204, 94-101 (2016-03-19)
Policosanol (PC) is a mixture of health promoting bioactive long-chain aliphatic alcohols. Here, we report that green tea (Camellia sinensis) leaves are the exceptionally rich plant-sources of PC. Young and tender leaves and old and turf leaves of C. sinensis
Ji-Eun Ra et al.
Food chemistry, 317, 126388-126388 (2020-02-23)
Policosanols is a health promoting aliphatic alcohol known as lipid-lowing agent. To enable maximising the functional properties of wheat, this research investigates the policosanol profiles and adenosine 5'-monophosphate-activated protein kinase (AMPK) activation potential of Korean wheat seedlings according to cultivars
Muthusamy Muthusamy et al.
Plants (Basel, Switzerland), 9(11) (2020-11-12)
The spectral quality and intensity of light, photoperiodism, and other environmental factors have profound impacts on the metabolic composition of light-dependent higher plants. Hence, we investigate the effects of fluorescent light (96 μmol m-2s-1) and white (100 μmol m-2s-1), blue
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service