Skip to Content
Merck
All Photos(1)

Key Documents

H6512

Sigma-Aldrich

Heparinase II from Flavobacterium heparinum

Lyophilized powder stabilized with approx. 25% bovine serum albumin, lyophilized powder, ≥100 units/mg protein (enzyme + BSA)

Synonym(s):

Heparin lyase II

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bacterial (Flavobacterium heparinum)

Quality Level

conjugate

conjugate (Glucosaminoglycan)

form

lyophilized powder

specific activity

≥100 units/mg protein (enzyme + BSA)

mol wt

84.1 kDa

storage temp.

−20°C

General description

Heparinase II is one of three key heparin-degrading enzymes of Flavobacterium heparinum (also known as Pedobacter heparinus1). It belongs to the polysaccharide lyases family PL21.

Application

Heparinase II from Flavobacterium heparinum has been used:
  • for digestion of heparin sulfate during exosome isolation
  • for digestion of heparin sulfate in notochordal cell conditioned media (NCCM) to investigate the content of glycosaminoglycans in NCCM
  • as a component of digestion buffer during cell surface glycan processing
  • as a component of heparin lyase (HSase) mix to remove the heparin sulfate on the 293ͰT/ACE2 cell surface and study the effect of histones on the infectivity of pseudovirus

Biochem/physiol Actions

Heparinase II cleaves heparan sulfate, and to a lesser extent heparin (relative activity about 2:1), at the α (1-4) linkages between hexosamines and uronic acid residues (both glucuronic and iduronic), yielding mainly disaccharides. Heparinase II has the broadest substrate specificity of the three heparinases Heparinase enzymes helps in decoding the complex structures of substrates. Heparin interferes with DNA transcription in PCR and in reverse transcription of RNA. Heparinase II has been used to remove heparin for downstream analysis of genomic DNA.

Unit Definition

One unit will form 0.1 μmole of unsaturated uronic acid per hr at pH 7.0 at 25 °C. One International Unit (I.U.) is equivalent to approx. 600 Sigma units.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

McLean, M.W., et al.
Proc. 8th Int. Symp. Glycoconjugates, 1, 73-73 (1985)
Moffat, C.F., et al.
Proc. 8th Int. Symp. Glycoconjugates, 1, 79-79 (1985)
PCR-sequence-specific primer typing of HLA class I and class II alleles.
Mike Bunce
Methods in molecular biology (Clifton, N.J.), 210, 143-171 (2002-11-05)
David Shaya et al.
The Journal of biological chemistry, 285(26), 20051-20061 (2010-04-21)
Heparinase II (HepII) is an 85-kDa dimeric enzyme that depolymerizes both heparin and heparan sulfate glycosaminoglycans through a beta-elimination mechanism. Recently, we determined the crystal structure of HepII from Pedobacter heparinus (previously known as Flavobacterium heparinum) in complex with a
Devina Purmessur et al.
The spine journal : official journal of the North American Spine Society, 15(5), 1060-1069 (2015-02-11)
Painful human intervertebral discs (IVDs) exhibit nerve growth deep into the IVD. Current treatments for discogenic back pain do not address the underlying mechanisms propagating pain and are often highly invasive or only offer temporary symptom relief. The notochord produces

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service