Skip to Content
Merck
All Photos(4)

Key Documents

A1651

Sigma-Aldrich

Adenosine 3′-phosphate 5′-phosphosulfate lithium salt hydrate

≥60%

Synonym(s):

3′-Phosphoadenosine 5′-phosphosulfate lithium salt, APPS, Active sulfate, PAPS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N5O13P2S · xLi+ · yH2O
CAS Number:
Molecular Weight:
507.26 (free acid basis)
Beilstein:
8181191
MDL number:
UNSPSC Code:
41106305
eCl@ss:
32160414
NACRES:
NA.51

form

powder

Quality Level

concentration

≥60%

shipped in

dry ice

storage temp.

−70°C

SMILES string

[Li+].[Li+].[Li+].[Li+].[P](=O)([O-])([O-])O[C@@H]1[C@H](O[C@H]([C@@H]1O)[n]2c3ncnc(c3nc2)N)CO[P](=O)([O-])O[S](=O)(=O)[O-].O

InChI

1S/C10H15N5O13P2S.4Li.H2O/c11-8-5-9(13-2-12-8)15(3-14-5)10-6(16)7(27-29(17,18)19)4(26-10)1-25-30(20,21)28-31(22,23)24;;;;;/h2-4,6-7,10,16H,1H2,(H,20,21)(H2,11,12,13)(H2,17,18,19)(H,22,23,24);;;;;1H2/q;4*+1;/p-4/t4-,6-,7-,10-;;;;;/m1...../s1

InChI key

UGODCLHJOJPPHP-AZGWGOJFSA-J

Looking for similar products? Visit Product Comparison Guide

Application

3′-Phosphoadenosine 5′-phosphosulfate (PAPS) is used as a universal sulfonate donor for in vivo sulfonation by sulfotransferases. PAPS is used for the sulfation of glycans. PAPS is also useful to study the distribution and function of its biological transporter(s).
Adenosine 3′-phosphate 5′-phosphosulfate lithium salt has been used in a study that cloned and characterized a mammalian lithium-sensitive bisphosphate 3′-nucleotidase inhibited by inositol 1,4-bisphosphate. Adenosine 3′-phosphate 5′-phosphosulfate lithium salt has also been used in a study to assess the golgi-resident PAP-specific 3′-phosphatase-coupled sulfotransferase assays.

Quality

Major impurities appear to be adenosine 3′,5′-diphosphate, sulfate and ammonia substitution products.

Caution

Rapid decomposition occurs when exposed to room temperature: up to 17% per day at 37 °C. We ship on dry ice to minimize decomposition in transit.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cooper, B.P. and Baumgarten, F.E.
Zeitschrift fur Naturforschung, 159 (1980)
Emanuel Smeds et al.
Glycobiology, 20(10), 1274-1282 (2010-06-18)
The signaling of various molecules involved in development and regulation of cell growth are regulated by heparan sulfate (HS). Specific binding of HS to ligand proteins depends on the HS sulfation pattern, where the spacing and number of O-sulfate groups
Paul A Dawson
Seminars in cell & developmental biology, 22(6), 653-659 (2011-03-23)
Sulfate (SO(4)(2-)) is an important nutrient for human growth and development, and is obtained from the diet and the intra-cellular metabolism of sulfur-containing amino acids, including methionine and cysteine. During pregnancy, fetal tissues have a limited capacity to produce sulfate
Xianxuan Zhou et al.
Glycobiology, 21(6), 771-780 (2011-01-13)
Heparan sulfate (HS) belongs to a major class of glycans that perform central physiological functions. Heparin is a specialized form of HS and is a clinically used anticoagulant drug. Heparin is a natural product isolated from pig intestine. There is
Saveria Pastore et al.
The Journal of investigative dermatology, 127(3), 660-667 (2006-10-14)
ATP is abundantly released from stressed or damaged cells in response to mechanical stimulation, bacteria, or noxious agents. In this study, we have investigated the possible involvement of P2 receptors (receptor for extracellular nucleotides) in the expression and release of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service