Skip to Content
Merck
All Photos(2)

Documents

C0954

Sigma-Aldrich

Chondroitinase C from Flavobacterium heparinum

lyophilized powder, ≥200 units/mg solid

Synonym(s):

Chondroitin C lyase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

conjugate

(Glucosaminoglycan)

Quality Level

form

lyophilized powder

specific activity

≥200 units/mg solid

storage temp.

−20°C

General description

Enzyme Commission (EC) Number 4.2.2.x

Biochem/physiol Actions

Chondroitinase C acts upon chondroitin sulfate C, releasing unsaturated 6-sulfated disaccharides. The enzyme also acts on hyaluronic acid, producing unsaturated nonsulfated disaccharides. The enzyme is strongly inhibited by NaCl, Fe2+, Co2+ and phosphate ions.

Unit Definition

One unit will form 0.1 μmole of unsaturated uronic acid per hr at pH 8.0 at 25 °C using chondroitin sulfate C as substrate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Ototani et al.
Carbohydrate research, 70(2), 295-306 (1979-05-01)
Chondroitinase B and chondroitinase C were separated from an extract of Flavobacterium heparinum induced with chondroitin 6-sulfate by using column chromatography on hydroxylapatite. Chondroitinase C was eluted together with the activities of hyaluronidase, delta4,5glycosiduronase, and sulfatase. The latter two activities
Y M Michelacci et al.
The Journal of biological chemistry, 251(4), 1154-1158 (1976-02-25)
A chondroitinase that acts upon chondroitin sulfate C and hyaluronic acid was isolated from Flavobacterium heparinum. This enzyme was seperated from constitutional chondroitinase AC and an induced chondroitinase B also present in extracts of F. heparinum previously grown in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service