Skip to Content
Merck
All Photos(1)

Key Documents

H4890

Sigma-Aldrich

Anti-Human Hemoglobin antibody produced in rabbit

whole antiserum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

human

technique(s)

indirect ELISA: 1:8,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HBB(3043)

General description

HBB (hemoglobin subunit β) codes for a β globin that consists of three exons. This gene is located on human chromosome 11p15. HBB is a serum protein that belongs to the histone-like protein family.

Immunogen

Hemoglobin isolated from normal adult human red blood cells and treated to remove lipoproteins.

Application

Anti-Human Hemoglobin antibody has been used in pulse chase experiments and western blotting.
Anti-Human Hemoglobin antibody produced in rabbit was used for immunoblotting using crude human cerebellar protein extracts and membrane extracts of human RBC. It was used for immunohistochemistry of human tongue cancer samples.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

HBB (hemoglobin subunit β) participates in oxygen transport from the lung to various peripheral tissues. Mutation in HBB result in Sickle cell disease (SCD). Overexpression of HBB has been observed in patients with periodontal disease.
Hemoglobin (Hb) is oxygen-binding metalloprotein that is present in the red blood cells. The most abundant Hb is HbA tetramer that is made up of two α, two β chains and the nonprotein heme group. The heme group contains iron ion and a porphyrin ring. Mutations in Hb gene result in hemoglobinopathies, the best known being sickle-cell disease and thalassemias.

Preparation Note

treated to remove lipoproteins

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amritha Suresh et al.
Disease markers, 32(1), 51-64 (2012-02-03)
Worldwide, the incidence of oral tongue cancer is on the rise, adding to the existing burden due to prevailing low survival and high recurrence rates. This study uses high-throughput expression profiling to identify candidate markers of resistance/response in patients with
A phylogenetic analysis of Borrelia burgdorferi sensu lato based on sequence information from the hbb gene, coding for a histone-like protein
Valsangiacomo C, et al.
International Journal of Systematic and Evolutionary Microbiology, 47(1), 1-10 (1997)
Proteomic analysis of saliva identifies potential biomarkers for orthodontic tooth movement
Ellias M F, et al.
TheScientificWorldJournal, 2012(1) (2012)
Manu Vanaerschot et al.
Nature microbiology, 2(10), 1403-1414 (2017-08-16)
Antimalarial compounds with dual therapeutic and transmission-blocking activity are desired as high-value partners for combination therapies. Here, we report the identification and characterization of hexahydroquinolines (HHQs) that show low nanomolar potency against both pathogenic and transmissible intra-erythrocytic forms of the
Abdullah Sultan et al.
The Journal of biological chemistry, 288(45), 32326-32342 (2013-10-01)
Fibril formation of β2-microglobulin and associated inflammation occur in patients on long term dialysis. We show that the plasma protein haptoglobin prevents the fatty acid-promoted de novo fibril formation of β2-microglobulin even at substoichiometric concentration. The fibrils are cytotoxic, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service