Skip to Content
Merck
All Photos(2)

Key Documents

WH0003043M1

Sigma-Aldrich

Monoclonal Anti-HBB antibody produced in mouse

clone 2H3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CD113tC, Anti-HBD, Anti-hemoglobin, Anti-hemoglobin, beta

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2H3, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

ELISA: suitable
capture ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG3κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HBB(3043)

General description

HBB (hemoglobin subunit β) codes for a β-globin that consists of three exons. This gene is located on human chromosome 11p15. HBB is a serum protein that belongs to the histone-like protein family.
The alpha (HBA) and beta (HBB) loci determine the structure of the 2 types of polypeptide chains in adult hemoglobin, Hb A. The normal adult hemoglobin tetramer consists of two alpha chains and two beta chains. Mutant beta globin causes sickle cell anemia. Absence of beta chain causes beta-zero-thalassemia. Reduced amounts of detectable beta globin causes beta-plus-thalassemia. The order of the genes in the beta-globin cluster is 5′-epsilon -- gamma-G -- gamma-A -- delta -- beta--3′. (provided by RefSeq)

Immunogen

HBB (AAH07075, 38 a.a. ~ 147 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
WTQRFFESFGDLSTPDAVMGNPKVKAHGKKVLGAFSDGLAHLDNLKGTFATLSELHCDKLHVDPENFRLLGNVLVCVLAHHFGKEFTPPVQAAYQKVVAGVANALPHKYH

Biochem/physiol Actions

HBB (hemoglobin subunit β) participates in oxygen transport from the lung to various peripheral tissues. Mutation in HBB result in Sickle cell disease (SCD). Overexpression of HBB has been observed in patients with periodontal disease.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proteomic analysis of saliva identifies potential biomarkers for orthodontic tooth movement
Ellias MF, et al.
TheScientificWorldJournal (2012)
A phylogenetic analysis of Borrelia burgdorferi sensu lato based on sequence information from the hbb gene, coding for a histone-like protein
Valsangiacomo C, et al.
International Journal of Systematic Bacteriology, 47(1), 1-10 (1997)
Loss of heterozygosity for chromosome 11 in primary human breast tumors is associated with poor survival after metastasis
Winqvist R, et al.
Cancer Research, 55(12), 2660-2664 (1995)
Two novel C-terminal frameshift mutations in the ?-globin gene lead to rapid mRNA decay
Rawa K, et al.
BMC Medical Genetics, 18(1), 65-65 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service