H1637
cis-4-Hydroxy-L-proline
collagen synthesis inhibitor
Synonym(s):
(2S,4S)-(−)-4-Hydroxy-2-pyrrolidinecarboxylic acid, CHP
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H9NO3
CAS Number:
Molecular Weight:
131.13
Beilstein:
81440
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Quality Level
Assay
≥98% (TLC)
form
powder
technique(s)
ligand binding assay: suitable
color
white to faint yellow
mp
257 °C (dec.) (lit.)
application(s)
peptide synthesis
SMILES string
O[C@@H]1CN[C@@H](C1)C(O)=O
InChI
1S/C5H9NO3/c7-3-1-4(5(8)9)6-2-3/h3-4,6-7H,1-2H2,(H,8,9)/t3-,4-/m0/s1
InChI key
PMMYEEVYMWASQN-IMJSIDKUSA-N
Looking for similar products? Visit Product Comparison Guide
Application
cis-4-Hydroxy-L-proline (CHP) is a proline analog that inhibits collagen synthesis and has been used as an anticancer compound. CHP blocked myotube formation and expression of sarcomeric myosin heavy chain in C2C12 murine skeletal muscle cells. CHP inhibited proliferation of murine Panc02 pancreatic carcinoma cell line and rat pancreatic carcinoma cell line DSL6A.
Biochem/physiol Actions
cis-4-Hydroxy-L-proline (CHP) and its derivatives may have anticancer activity. cis-4-Hydroxy-L-proline and other hydrosyl-L-prolines are important intermediates for chiral synthesis of potential drugs.
Other Notes
Constituent of Amanita sp. peptide toxins but not of animal proteins like collagen.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Christoph Mueller et al.
World journal of gastroenterology, 12(10), 1569-1576 (2006-03-30)
To investigate the biological effects of cis-hydroxyproline (CHP) on the rat pancreatic carcinoma cell line DSL6A, and to examine the underlying molecular mechanisms. The effect of CHP on DSL6A cell proliferation was assessed by using BrdU incorporation. The expression of
Heather Dickens et al.
Molecular medicine reports, 1(4), 459-464 (2008-07-01)
cis-4-Hydroxy-L-proline (CHP) is being clinically evaluated as an anticancer drug. Since this compound targets the production of L-proline-rich proteins and critical L-proline residues, its impact on long-term cultures of human hepatocytes and toxicity in rats was studied to investigate possible
Dietrich Sturm et al.
International journal of colorectal disease, 25(8), 921-929 (2010-04-21)
This study addressed the question of whether the collagen metabolism modulator cis-4-Hydroxy-L-proline (CHP) is applicable for potential use as a therapeutic inhibitor of pancreatic carcinoma cell growth. Cell proliferation, as well as quantification of apoptosis of murine Panc02 cells, was
Linda Metzner et al.
The Journal of pharmacology and experimental therapeutics, 309(1), 28-35 (2004-01-14)
Several proline derivatives such as L-azetidine-2-carboxylic acid, cis-4-hydroxy-L-proline, and 3,4-dehydro-DL-proline prevent procollagen from folding into a stable triple-helical conformation, thereby reducing excessive deposition of collagen in fibrotic processes and the growth of tumors. This study was performed to investigate whether
Shin-Ichiro Kurimoto et al.
Chemistry & biodiversity, 12(8), 1200-1207 (2015-08-13)
Coleifolides A and B (1 and 2), two new sesterterpenoids with a β-methyl-α,β-unsaturated-γ-lactone moiety, were isolated from the aerial parts of Scutellaria coleifolia H.Lév. (Lamiaceae), together with three known compounds. Their structures were elucidated by NMR and MS examinations. Coleifolides
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service