GE17-1353-01
HiTrap® Phenyl Fast Flow (lS)
Cytiva 17-1353-01, pack of 5 × 1 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
23151816
NACRES:
NA.56
Recommended Products
ligand
phenyl
packaging
pack of 5 × 1 mL
manufacturer/tradename
Cytiva 17-1353-01
parameter
<4 mL/min flow rate
42 psi
bed size
7 mm × 25 mm
bed volume
1 mL
column I.D.
7 mm
matrix
6% cross-linked agarose
particle size
45-165 μm
average diameter
90 μm
cleaning
2-14
working range
3-13
suitability
suitable for bioprocess medium
General description
HiTrap® Phenyl FF (Low Sub) are prepacked Phenyl Sepharose™ 6 Fast FLow columns for hydrophobic interaction chromatography (HIC). The columns are used for small-scale protein purification as well as for screening of binding and elution conditions.
Features and Benefits
- Provide convenient and fast small-scale Protein purifications.
- Phenyl Sepharose™ 6 Fast FLow is a well-established, standard aromatic HIC medium for capture and intermediate purification requiring Low to medium hydrophobicity. Two levels of substitution on Phenyl Sepharose™ 6 Fast FLow help find optimal selectivity and binding capacity.
- Flexible design for use with syringes, single laboratory pump, or chromatography system such as ÄKTA® design.
- High flow rates and high capacities.
- Excellent and easy scale-up achieved by connecting several HiTrap® columns in series or using HiPrep™ columns.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Other Notes
The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.
Legal Information
HiPrep is a trademark of Cytiva
HiTrap is a registered trademark of Cytiva
Sepharose is a trademark of Cytiva
ÄKTA is a registered trademark of Cytiva
related product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service