Skip to Content
Merck
All Photos(1)

Key Documents

A50120

Sigma-Aldrich

DEAE–Sephadex®

Synonym(s):

Diethylaminoethyl Sephadex®

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
47101511
NACRES:
NA.56
Pricing and availability is not currently available.

form

powder

Quality Level

technique(s)

protein purification: suitable

matrix

cross-linked dextran

bead size

40-125 μm (dry)

pore size

~200,000 Da exclusion limit

pH

2—12

capacity

3-4 meq/g binding capacity

General description

A50120-100G′s updated product number is GE17-0180-01
A50120-500G′s updated product number is GE17-0180-02

Application

DEAE Sephadex can be used in ion exchange chromatography for purifying and isolating proteins. In has been used to identify and label an insulin activated nitric oxide synthase inhibitor (protein) in acute myocardial infarction (AMI) patients.

Legal Information

Sephadex is a registered trademark of Cytiva

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Okuno et al.
Biochimica et biophysica acta, 841(1), 112-119 (1985-07-26)
Quinolinic acid phosphoribosyltransferase (EC 2.4.2.19) was purified 3600-fold from rat liver and 280-fold from rat brain. Kinetic analyses (Km = 12 microM for the substrate quinolinic acid and Km = 23 microM for the cosubstrate phosphoribosylpyrophosphate), physicochemical properties of the
Clovis R Nakaie et al.
Analytical biochemistry, 318(1), 39-46 (2003-06-05)
This report demonstrates that due to the presence of residual reactive sites in their matrices, classical diethylaminoethyl-attaching commercial anion-exchanger resins such as DEAE-MacroPrep and DEAE-Sephadex A50 supports can be used for peptide synthesis. Moreover, due to the high stability of
Misook Kim et al.
Biochimica et biophysica acta, 1770(12), 1627-1635 (2007-10-09)
Two cysteine proteases, GP2 and GP3, have been isolated from ginger rhizomes (Zingiber officinale). GP2 is virtually identical to a previously identified ginger protease GPII [K.H. Choi, and R.A. Laursen, Amino-acid sequence and glycan structures of cysteine proteases with proline
P K Farmer et al.
Biochimica et biophysica acta, 1434(1), 6-17 (1999-11-11)
A calmodulin-like domain protein kinase (DcCPK1, previously designated CDPK431) cloned from carrot (Daucus carota L.) was expressed at high levels in Escherichia coli and partially purified. Ca(2+)-induced gel mobility shift and (45)Ca(2+) ligand binding assays confirmed that recombinant DcCPK1 binds
M A Zaval'nyĭ et al.
Voprosy virusologii, (5)(5), 583-589 (1980-09-01)
Primary cells of chick embryos (CEC), new-born piglet kidneys (NPK), and green monkey kidney (GMK) obtained by the conventional method of tissue trypsinization were shown to grow satisfactorily on microcarriers of various types. CEC formed a confluent monolayer on Supperbead

Questions

1–5 of 5 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. How can Product A50120, DEAE-Sephadex® resin be regenerated?

    1 answer
    1. Regeneration is generally accomplished by washing with high ionic strength buffer or decreasing/increasing pH. The resin is then re-equilibrated in binding buffer.

      Helpful?

  3. What is the procedure for swelling Product A50120, DEAE-Sephadex® resin?

    1 answer
    1. Weigh out the required amount of dry powder and suspend it in the buffer of choice. Although the swelling volume will be specific to the binding buffer used, generally 1 gram of powder will swell to 25 mL final volume of medium. Excess binding buffer should be used. Swell the resin for 1-2 days at room temperature or 2 hours at 100°C over boiling water. After swelling, wash the resin on a Buchner funnel with binding buffer. Prepare a slurry with 75% swelled medium/25% binding buffer.

      Helpful?

  4. What procedures are used for elution when using Product A50120, DEAE-Sephadex® resin?

    1 answer
    1. For DEAE Sephadex, elution is achieved using either an increasing salt gradient or a decreasing pH gradient.

      Helpful?

  5. How can Product A50120, DEAE-Sephadex® resin, be stored?

    1 answer
    1. Store swollen medium refrigerated in the presence of a preservative such as 20% Ethanol. Sodium azide or thimerosal should not be used.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service