GE10600087
Amersham™ Hybond® P Western blotting membranes, PVDF
pore size 0.45 μm, sheet W × L 100 mm × 100 mm, pkg of 10 ea
All Photos(1)
About This Item
UNSPSC Code:
41105339
NACRES:
NB.22
Recommended Products
packaging
pkg of 10 ea
manufacturer/tradename
Cytiva 10600087
sheet W × L
100 mm × 100 mm
pore size
0.45 μm pore size
Related Categories
General description
PVDF membranes have higher mechanical strength than NC membranes and, thus, are more suitable for Western blotting applications where stripping and reprobing is needed. In addition, PVDF membranes exhibit higher protein binding capacity, however, with a tendency for a higher background, compared with NC membranes.
Features and Benefits
- Protein binding capacity over 200 µg/cm2 for easy signal detection
- High protein retention even after harsh washing steps.
- High protein capture during transfer minimizes sample loss.
- Available in popular precut sizes for your application.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Legal Information
Amersham is a trademark of Cytiva
Hybond is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)