Skip to Content
Merck
All Photos(1)

Key Documents

GE10600021

Amersham Hybond® P Western blotting membranes, PVDF

pore size 0.2 μm, roll W × L 260 mm × 4 m, pkg of 1 ea

Synonym(s):

PVDF blotting membrane, Western blot membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
NACRES:
NB.22

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 10600021

roll W × L

260 mm × 4 m

pore size

0.2 μm pore size

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

PVDF membranes have higher mechanical strength than NC membranes and, thus, are more suitable for Western blotting applications where stripping and reprobing is needed. In addition, PVDF membranes exhibit higher protein binding capacity, however, with a tendency for a higher background, compared with NC membranes.

Features and Benefits

  • Easy signal detection
  • Offers high protein retention even after harsh washing steps and is available in common precut sizes
  • Captures maximum proteins during transfers aids in minimizing sample loss
  • The pore size of 0.2 μm for increased surface area, resulting in enhanced binding of low molecular weight proteins
  • For use with standard colorimetric and chemiluminescent detection methods

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

Amersham is a trademark of Cytiva
Hybond is a registered trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service