Skip to Content
Merck
All Photos(1)

Key Documents

IPVH00010

Millipore

Immobilon® -P PVDF Membrane

1 roll, 27 cm x 3.75 m, 0.45 µm pore size, transfer membrane

Synonym(s):

Western blotting membrane, blotting membrane, transfer membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
eCl@ss:
32031602
NACRES:
NB.22

Product Name

Immobilon®-P PVDF Membrane, 1 roll, 27 cm x 3.75 m, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.

material

PVDF membrane
plain filter
white filter

Quality Level

feature

hydrophobic

manufacturer/tradename

Immobilon®

technique(s)

dot blot: suitable
western blot: suitable

filter L × W

27 cm × 3.75 m

pore size

0.45 μm pore size

capacity

160 μg/cm2 adsorption capacity (insulin)
215 μg/cm2 adsorption capacity (BSA)
294 μg/cm2 adsorption capacity (goat IgG)

compatibility

for use with Amido black
for use with CPTS
for use with Colloidal gold
for use with Coomassie brilliant blue
for use with India ink
for use with Ponceau-S red
for use with Sypro<TMSYMBOL></TMSYMBOL> ruby
for use with Toluidine blue
for use with Transillumination

detection method

chemiluminescent
colorimetric
fluorometric
radioactive

shipped in

ambient

General description

Immobilon-P transfer membrane is recommended for most Western blotting applications, especially proteins greater than 20 kDa. Immobilon-P membrane has excellent protein retention, high physical strength, and broad chemical compatibility, making it ideal for a variety of staining applications and reprobing in immunodetection methods.

Features and Benefits

Immobilon-P PVDF Membrane:
  • Won′t crack, curl or fracture when cut
  • Low background
  • Superior staining capabilities
  • Can be reprobed multiple times

Linkage

Replaces: INCP00010

Other Notes

Filter Code: IPVH
Primary Binding Mechanisms: Electrostatic, hydrophobic

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabelle Robert et al.
The Journal of experimental medicine, 206(5), 1047-1056 (2009-04-15)
Immunoglobulin class switch recombination (CSR) is initiated by DNA breaks triggered by activation-induced cytidine deaminase (AID). These breaks activate DNA damage response proteins to promote appropriate repair and long-range recombination. Aberrant processing of these breaks, however, results in decreased CSR
Effects of HIV protease inhibitors on progression of monocrotaline- and hypoxia-induced pulmonary hypertension in rats.
Gary-Bobo, Guillaume, et al.
Circulation, 122, 1937-1947 (2010)
Remodeling of VE-cadherin junctions by the human herpes virus 8 G-protein coupled receptor.
Dwyer, J, et al.
Oncogene, 30, 190-200 (2011)
Interactions between Brucella suis VirB8 and its homolog TraJ from the plasmid pSB102 underline the dynamic nature of type IV secretion systems.
Bourg, Gisele, et al.
Journal of Bacteriology, 191, 2985-2992 (2009)
Pulmonary artery smooth muscle cell senescence is a pathogenic mechanism for pulmonary hypertension in chronic lung disease.
Noureddine, Hibo, et al.
Circulation Research, 109, 543-553 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service