Skip to Content
Merck
All Photos(1)

Key Documents

G9910

Sigma-Aldrich

Sodium glycodeoxycholate

BioXtra, ≥97% (HPLC)

Synonym(s):

3α,12α-Dihydroxy-5β-cholan-24-oic acid N-(carboxymethyl)amide, Glycodeoxycholic acid sodium salt, Glycodesoxycholic acid, N-(3α,12α-Dihydroxy-24-oxocholan-24-yl)glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H42NNaO5
CAS Number:
Molecular Weight:
471.61
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

anionic

Quality Level

product line

BioXtra

Assay

≥97% (HPLC)

mol wt

471.61 g/mol

impurities

≤0.0025% Phosphorus (P)
≤0.1% Insoluble matter

CMC

2.1 mM

solubility

H2O: 0.1 M, clear to slightly hazy, colorless to almost colorless

anion traces

chloride (Cl-): ≤1%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.011%
Cu: ≤0.0005%
Fe: ≤0.001%
K: ≤0.005%
NH4+: ≤0.05%
Pb: ≤0.001%
Zn: ≤0.003%

SMILES string

[Na].[H][C@]12CC[C@@]3([H])[C@]4([H])CC[C@]([H])([C@H](C)CCC(=O)NCC(O)=O)[C@@]4(C)[C@@H](O)C[C@]3([H])[C@@]1(C)CC[C@@H](O)C2

InChI

1S/C26H43NO5.Na.H/c1-15(4-9-23(30)27-14-24(31)32)19-7-8-20-18-6-5-16-12-17(28)10-11-25(16,2)21(18)13-22(29)26(19,20)3;;/h15-22,28-29H,4-14H2,1-3H3,(H,27,30)(H,31,32);;/t15-,16-,17-,18+,19-,20+,21+,22+,25+,26-;;/m1../s1

InChI key

BMYWWGPEKRLDGW-ZXMGEDLUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium glycodeoxycholate is a bile salt that can be used to form biomimicking environments.

Application

Sodium glycodeoxycholate has been used in a study to develop more similar mimetic intestinal fluids to physiological fluids than those developed by mixed bile salt systems (MBS). It has also been used in a study to investigate Ca2+ buffering capability of bile salt micelles.

Biochem/physiol Actions

Induces apoptosis in hepatocytes; may induce DNA cleavage.

Other Notes

Bile Salt

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bile salt composition is secondary to bile salt concentration in determining hydrocortisone and progesterone solubility in intestinal mimetic fluids
Huda H Zughaid, et al.
International Journal of Pharmaceutics, 422, 7-7 (2012)
Domenico Trombetta et al.
Nutrients, 12(11) (2020-11-11)
Almond kernels contain phytochemicals with positive health effects in relation to heart disease, diabetes and obesity. Several studies have previously highlighted that almond cell wall encapsulation during digestion and particle size are factors associated with these benefits. In the present
Hu-He Chen et al.
Guang pu xue yu guang pu fen xi = Guang pu, 30(3), 748-752 (2010-05-26)
In the present article, NaGDC was used to study the Ca(2+)-buffering capability of bile salt micelles. NaGDC is a naturally occurring bile salt which constitutes approximately 10 molar percent of bile salt composition in gallbladder. The authors selected glycine conjugated
Raina Thakur et al.
Photochemistry and photobiology, 88(5), 1248-1255 (2012-05-23)
Photophysical properties of two widely used antibiotic fluoroquinolone drugs, namely Norfloxacin (NOR) and Ofloxacin (OFL) have been investigated in biomimicking environments formed by bile salts. Experimental results demonstrate that photophysical enhancement and fall of a particular prototropic species are sensitive
Laura M Sanchez et al.
PloS one, 11(3), e0149603-e0149603 (2016-03-19)
Biofilms are a ubiquitous feature of microbial community structure in both natural and host environments; they enhance transmission and infectivity of pathogens and provide protection from human defense mechanisms and antibiotics. However, few natural products are known that impact biofilm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service