Skip to Content
Merck
All Photos(6)

Documents

T4009

Sigma-Aldrich

Taurocholic acid sodium salt hydrate

≥95% (HPLC)

Synonym(s):

2-[(3α,7α,12α-Trihydroxy-24-oxo-5β-cholan-24-yl)amino]ethanesulfonic acid, 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid N-(2-sulfoethyl)amide, Sodium taurocholate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H44NNaO7S · xH2O
CAS Number:
Molecular Weight:
537.68 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

description

anionic

Assay

≥95% (HPLC)

form

powder

mol wt

micellar avg mol wt 2100

aggregation number

4

CMC

3-11 mM (20-25°C)

functional group

sulfonic acid

shipped in

ambient

storage temp.

room temp

SMILES string

[Na+].[H]O[H].[H][C@@]12C[C@H](O)CC[C@]1(C)[C@@]3([H])C[C@H](O)[C@]4(C)[C@H](CC[C@@]4([H])[C@]3([H])[C@H](O)C2)[C@H](C)CCC(=O)NCCS([O-])(=O)=O

InChI

1S/C26H45NO7S.Na.H2O/c1-15(4-7-23(31)27-10-11-35(32,33)34)18-5-6-19-24-20(14-22(30)26(18,19)3)25(2)9-8-17(28)12-16(25)13-21(24)29;;/h15-22,24,28-30H,4-14H2,1-3H3,(H,27,31)(H,32,33,34);;1H2/q;+1;/p-1/t15-,16+,17-,18-,19+,20+,21-,22+,24+,25+,26-;;/m1../s1

InChI key

RDAJAQDLEFHVNR-NEMAEHQESA-M

Looking for similar products? Visit Product Comparison Guide

General description

Taurocholic acid sodium salt hydrate is a bile acid with detergent capabilities.

Application

Taurocholic acid sodium salt hydrate has been used in a study to assess the adsorption of bile acids and salts to vegetable fibrous tissue. It has also been used in a study to investigate a method for determination of conjugated and unconjugated bile salts in serum and jejunal fluid.
Anionic detergent used for protein solubilization.

Biochem/physiol Actions

Non-sulfated bile salt, physiological transport substrate for the bile salt export pump/sister of Pgp (BSEP/spgp), Na(+)/taurocholate cotransporter (NTCP) and MRP3 into the hepatic carnalicular.

Preparation Note

Synthesized from cholic acid

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Studies on the adsorption of bile salts to non-absorbed components of diet.
M A Eastwood et al.
Biochimica et biophysica acta, 152(1), 165-173 (1968-01-10)
Studies on the Antibacterial Properties of the Bile Acids and some Compounds Derived from Cholanic Acid
M. Stacey and M. Webb
Proc. Royal Soc. Lond. B., 134, 523-523 (1947)
Yanxia Wei et al.
BMC infectious diseases, 19(1), 46-46 (2019-01-13)
Clostridium difficile (C. difficile) is a main cause of antibiotic-associated diarrhoea in humans. Several studies have been performed to reveal the prevalence rate of C. difficile in cats and dogs. However, little is known about the epidemiology of C. difficile
Theresa D Ho et al.
Infection and immunity, 82(6), 2345-2355 (2014-03-26)
Clostridium difficile is a clinically important pathogen and the most common cause of hospital-acquired infectious diarrhea. Expression of the C. difficile gene csfV, which encodes σ(V), an extracytoplasmic function σ factor, is induced by lysozyme, which damages the peptidoglycan of
A Tangerman et al.
Clinica chimica acta; international journal of clinical chemistry, 159(2), 123-132 (1986-09-15)
A reliable method is described for the determination of conjugated and unconjugated bile acids in serum and jejunal fluid. Bile acids are extracted using reverse-phase octadecylsilane bonded silica cartridges and are separated into their unconjugated and conjugated fractions using the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service