Skip to Content
Merck
All Photos(4)

Key Documents

T0557

Sigma-Aldrich

Sodium taurodeoxycholate hydrate

BioXtra, ≥97% (TLC)

Synonym(s):

2-([3α,12α-Dihydroxy-24-oxo-5β-cholan-24-yl]amino)ethanesulfonic acid, Taurodeoxycholic acid sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C26H44NO6SNa · xH2O
CAS Number:
Molecular Weight:
521.69 (anhydrous basis)
Beilstein:
3901204
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

anionic

product line

BioXtra

Assay

≥97% (TLC)

mol wt

micellar avg mol wt 3100

aggregation number

6

impurities

≤0.002% Phosphorus (P)
≤0.1% Insoluble matter

CMC

1-4 mM (20-25°C)

solubility

H2O: 0.5 M, clear, colorless to faintly yellow

anion traces

chloride (Cl-): ≤0.5%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.01%
Mg: ≤0.005%
NH4+: ≤0.05%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

O.[Na+].C[C@H](CCC(=O)NCCS([O-])(=O)=O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

InChI

1S/C26H45NO6S.Na.H2O/c1-16(4-9-24(30)27-12-13-34(31,32)33)20-7-8-21-19-6-5-17-14-18(28)10-11-25(17,2)22(19)15-23(29)26(20,21)3;;/h16-23,28-29H,4-15H2,1-3H3,(H,27,30)(H,31,32,33);;1H2/q;+1;/p-1/t16-,17-,18-,19+,20-,21+,22+,23+,25+,26-;;/m1../s1

InChI key

OLPIZAYYAVQETM-GGPRKOIFSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium tauroursodeoxycholate is a bile salt which is naturally present in the small bowel.

Application

Bile salt-related, anionic detergent used for isolation of membrane proteins including inner mitochondrial membrane proteins.
Sodium tauroursodeoxycholate has been used in a study to assess short-term feedback regulation of bile salt uptake in rodent liver. It has also been used in a study to investigate its potential to resolve endoplasmic reticulum (ER) stress in intestinal epithelial cells. It is also used for isolation of membrane proteins including inner mitochondrial membrane proteins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emanuel Berger et al.
Biochemical and biophysical research communications, 409(4), 610-615 (2011-05-25)
Inflammatory bowel diseases (IBD) are chronically relapsing and immune-mediated disorders of the gastrointestinal tract. Endoplasmic reticulum (ER) stress mechanisms in the epithelium have been demonstrated to be implemented into the pathogenesis of intestinal inflammation. Chemical chaperones have been demonstrated to
Stefanie Mühlfeld et al.
Hepatology (Baltimore, Md.), 56(6), 2387-2397 (2012-07-19)
The sodium taurocholate cotransporting polypeptide (Ntcp) is the major bile salt uptake transporter at the sinusoidal membrane of hepatocytes. Short-term feedback regulation of Ntcp by primary bile salts has not yet been investigated in vivo. Subcellular localization of Ntcp was
Kalinne Kelly Lima Gadelha et al.
Experimental physiology, 104(2), 199-208 (2018-12-19)
What is the central question of this study? Is the responsiveness of isolated segments of the rat oesophagus to contractile or relaxant stimuli susceptible to acute luminal exposure of the oesophagus to an acid solution that contains pepsin and bile
Salma Alavi et al.
Cell, 181(7), 1533-1546 (2020-07-08)
The gut microbiome is the resident microbial community of the gastrointestinal tract. This community is highly diverse, but how microbial diversity confers resistance or susceptibility to intestinal pathogens is poorly understood. Using transplantation of human microbiomes into several animal models
Kai Zheng et al.
Molecular nutrition & food research, 64(14), e1901231-e1901231 (2020-06-02)
The development of novel compounds that trigger non-apoptotic cell death may represent alternative therapeutic strategies for esophageal squamous cell carcinoma (ESCC) treatment. Cellular senescence suppresses tumorigenesis by halting the proliferation of tumor cells, implying the induction of senescence as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service