Skip to Content
Merck
All Photos(1)

Key Documents

G9136

Sigma-Aldrich

Porcine Gelatin

from porcine skin, Type A, powder, γ-irradiated, suitable for cell culture, BioXtra

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.75

product name

Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture

biological source

Porcine skin

Quality Level

sterility

γ-irradiated

type

Type A

product line

BioXtra

form

lyophilized powder

packaging

pkg of 10 mg

technique(s)

cell culture | mammalian: suitable

surface coverage

100‑200 μg/cm2

gel strength

~300 g Bloom

solubility

soluble 20 mg/mL, clear, colorless (in warm tissue culture medium)

shipped in

ambient

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

This product is recommended for use as a cell culture substratum at 100-200 μg/cm2. The optimal concentration does depend on cell type as well as the application and research objectives.

Gelatin has been used in many applications. It has use in coating cell culture to improve attachment of cells, being added to PCR to stabilize Taq DNA, as a blocking reagent in Western blotting, ELISA, and immunochemistry, and as a component of media for species differentiation in bacteriology. As a biocompatible polymer, it has used as a delivery vehicle for release of active biomolecules and in generation of scaffolds for tissue engineering applications. In the pharmaceutical industry, geltan can be used as a suspending and encapsulating agent, among other applications.

Components

Gelatin is a heterogeneous mixture of water-soluble proteins of high average molecular masses, present in collagen. Proteins are extracted by boiling the relevant skin, tendons, ligaments, bones, etc. in water. Type A gelatin is derived from acid-cured tissue. Type B is derived from lime-cured tissue.

Caution

Dry gelatin, when stored in airtight containers at room temperature, will remain unchanged for many years. When heated at 100°C in the presence of air, it swells becomes soft and disintegrates to a carbonaceous mass with evolution of pyridine bases and ammonia.

Preparation Note

This product is derived from porcine skin. Gelatin is soluble in hot than in cold water. It is practically insoluble in most organic solvents such as alcohol, chloroform, carbon disulfide, carbon tetrachloride, ether, benzene, acetone, and oils. The Bloom number, determined by the Bloom gelometer, is an indication of the strength of a gel formed from a solution of the known concentration. The Bloom number is proportional to the average molecular mass. Bloom numbers of porcine skin Gelatin vary from 90 to 300 g.

This product is a type A lyophilized powder, suitable for cell culture, with a gel strength of 300.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonia Capellero et al.
International journal of molecular sciences, 23(2) (2022-01-22)
Peritoneal metastases are the leading cause of morbidity and mortality in ovarian cancer. Cancer cells float in peritoneal fluid, named ascites, together with a definitely higher number of non neo-neoplastic cells, as single cells or multicellular aggregates. The aim of
Giulia Villari et al.
The EMBO journal, 39(24), e103661-e103661 (2020-11-21)
Although subcellular positioning of endosomes significantly impacts on their functions, the molecular mechanisms governing the different steady-state distribution of early endosomes (EEs) and late endosomes (LEs)/lysosomes (LYs) in peripheral and perinuclear eukaryotic cell areas, respectively, are still unsolved. We unveil
Carla Luana Dinardo et al.
PloS one, 10(12), e0145062-e0145062 (2015-12-15)
Recent evidence shows the rigidity of vascular smooth muscle cells (VSMC) contributes to vascular mechanics. Arterial rigidity is an independent cardiovascular risk factor whose associated modifications in VSMC viscoelasticity have never been investigated. This study's objective was to evaluate if
Ewelina Rubin et al.
Diseases of aquatic organisms, 122(1), 21-33 (2016-12-03)
Quahog parasite unknown (QPX) is a protistan parasite capable of causing deadly infections in the hard clam Mercenaria mercenaria, one of the most valuable shellfish species in the USA. QPX is an extracellular parasite found mostly in the connective tissue
Marta Czernik et al.
Nature protocols, 11(11), 2170-2188 (2016-10-07)
This protocol describes how to convert the chromatin structure of sheep and mouse somatic cells into spermatid-like nuclei through the heterologous expression of the protamine 1 gene (Prm1). Furthermore, we also provide step-by-step instructions for somatic cell nuclear transfer (SCNT)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service