Skip to Content
Merck
All Photos(4)

Key Documents

G7127

Sigma-Aldrich

Guanosine 5′-diphosphate sodium salt

Guanosine 5′-diphosphate sodium salt

Type I, ≥96% (HPLC)

Synonym(s):

GDP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N5O11P2
CAS Number:
Molecular Weight:
443.20
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
Pricing and availability is not currently available.

biological source

synthetic (organic)

Quality Level

type

Type I

Assay

≥96% (HPLC)

form

powder

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na].NC1=NC(=O)c2ncn(C3OC(COP(O)(=O)OP(O)(O)=O)C(O)C3O)c2N1

InChI

1S/C10H15N5O11P2.Na.H/c11-10-13-7-4(8(18)14-10)12-2-15(7)9-6(17)5(16)3(25-9)1-24-28(22,23)26-27(19,20)21;;/h2-3,5-6,9,16-17H,1H2,(H,22,23)(H2,19,20,21)(H3,11,13,14,18);;

InChI key

PVMPTWQLNDHIEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Guanosine 5′-diphosphate sodium salt has been used:
  • in GTP-γ-35S functional assays[1]
  • in Y1 agonist-stimulated [35S]- guanosine-5′-O-(3-thio)triphosphate (GTPγS) functional binding[2]
  • in the expression and purification of recombinant proteins[3]

Biochem/physiol Actions

Guanosine 5′-diphosphate (GDP) is the diphosphate nucleoside of the purine guanine. GDP and GTP are involved in and may be used to study cell signaling via G-coupled protein receptors (GCPR) and guanine nucleotide exchange factors (GEF) and a variety of GTPases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N-desalkylquetiapine, a potent norepinephrine reuptake inhibitor and partial 5-HT 1A agonist, as a putative mediator of quetiapine's antidepressant activity
Jensen N H, et al.
Neuropsychopharmacology, 33(10), 2303-2303 (2008)
María-de-Los-Angeles Andrade-Oliva et al.
Environmental toxicology and pharmacology, 60, 58-65 (2018-04-17)
Male Sprague-Dawley rats (8-9 weeks-old) were exposed for three days (acute exposure) or eight weeks (subchronic exposure) to purified air or concentrated ambient fine particles, PM2.5 (≤2.5 μm; 15 to 18-fold of ambient air; 370-445 μg/m3). In membranes from rat prefrontal cortex
Neuropeptide Y promotes neurogenesis in murine subventricular zone
Agasse F, et al.
Stem Cells, 26(6), 1636-1645 (2008)
Nami Kim et al.
Scientific reports, 7, 41066-41066 (2017-01-25)
This study characterizes the human metabolic response to piperine, a curcumin extract, and the details of its underlying molecular mechanism. Using
A p120 catenin isoform switch affects Rho activity, induces tumor cell invasion, and predicts metastatic disease
Yanagisawa M, et al.
The Journal of Biological Chemistry, 283(26), 18344-18354 (2008)

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service