Skip to Content
Merck
All Photos(4)

Key Documents

G7781

Sigma-Aldrich

Anti-Glutathione-S-Transferase (GST) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Glutathione-S-Transferase, Anti GST Antibody - Anti-Glutathione-S-Transferase (GST) antibody produced in rabbit, Anti Gst Antibody, Anti-Gst Antibody, Gst Antibody, Anti-GST

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

packaging

antibody small pack of 25 μL

technique(s)

dot blot: suitable
indirect ELISA: 1:10,000
western blot: 1:2,000 using lysates of induced Escherichia coli expressing GST

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Anti-Glutathione-S-Transferase (GST) antibody produced in rabbit is specific for native as well as denatured-reduced forms of glutathione-S-transferase from Schistosoma japonicum. The soluble GST is encoded by eight different genes in humans namely, α, μ, θ, π, ζ, σ, κ and ω on chromosome 6, 1, 22, 11, 14, 4, not known and 10 respectively.
The antibody is specific for native as well as denatured-reduced forms of glutathione-S-transferase from Schistosoma japonicum. Anti-GST may be used in various immunoassays to identify the expression of GST fusion proteins.

Immunogen

recombinant GST from Schistosoma japonicum expressed in E. coli.

Application

Anti-Glutathione-S-Transferase (GST) antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

Glutathione-S-transferase (GST) catalyzes the detoxication of electrophiles by glutathione conjugation. This enzyme provides protection against oxidative stress and toxic foreign chemicals. Polymorphism in GST gene is associated with the development of asthma and cutaneous basal cell carcinoma.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glutathione-S-transferase family of enzymes
Strange R, et al.
Mutation Research. Fundamental and Molecular Mechanisms of Mutagenesis, 482(1-2), 21-26 (2001)
Analysis of protein-ligand interactions by fluorescence polarization
Rossi AM and TaylorCW
Nature Protocols, 6(3), 365-365 (2011)
Glutathione S-transferase polymorphisms and their biological consequences
Hayes JD and Strange RC
Pharmacology, 61(3), 154-166 (2000)
A novel simple extracellular leucine-rich repeat (eLRR) domain protein from rice (OsLRR1) enters the endosomal pathway and interacts with the hypersensitive-induced reaction protein 1 (OsHIR1).
Zhou L, et al.
Plant, Cell and Environment, 32(12), 1804-1820 (2009)
Nina Schroeder et al.
Journal of virology, 86(9), 4868-4882 (2012-02-22)
Mature vaccinia virus (vaccinia MV) infects a broad range of animals in vivo and cell cultures in vitro; however, the cellular receptors that determine vaccinia MV tropism and entry pathways are poorly characterized. Here, we performed quantitative proteomic analyses of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service