Skip to Content
Merck
All Photos(1)

Key Documents

GE27-4577-01

Sigma-Aldrich

Anti-GST Antibody

Cytiva 27-4577-01

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352116
NACRES:
NA.32

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

manufacturer/tradename

Cytiva 27-4577-01

packaging

pkg of 0.5 mL, 50 detections

shipped in

wet ice

storage temp.

2-8°C

Related Categories

General description

Anti-GST Antibody is a polyclonal antibody purified from the sera of goats immunized with purified schistosomal GST. Because of its polyclonal nature, it can recognize more than one epitope on GST, thereby improving its capacity for recognizing GST fusion proteins even if some binding sites are masked due to recombinant protein folding.

Application

Western Blots

Features and Benefits

  • For the highly sensitive and specific detection of recombinant schistosomal GST fusion proteins expressed using pGEX vectors
  • Extensively cross-adsorbed to remove antibodies that bind to E. coli proteins and affinity purified using Glutathione Sepharose
  • Supplied unconjugated for use with any enzyme-conjugated anti-goat antibody
  • Recommended for use in Western blots and dot blots to detect GST fusion proteins

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

GST Gene Fusion Vectors
A license for commercial use of GST Gene Fusion Vectors under US patent 5,654,176 and equivalent patents and patent applications in other countries must be obtained from Millipore Corp (formerly Chemicon International Inc).
Sepharose is a trademark of Cytiva

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazumi Nakano et al.
Molecular cancer research : MCR, 17(12), 2522-2536 (2019-10-09)
The genetic and molecular alterations responsible for leukemogenesis and progression of HTLV-infected adult T-cell leukemia (ATL) have not been fully clarified. Previously, we reported that various genes are not only overexpressed but also abnormally spliced in ATL cells. Here, we
Stefano Rosa et al.
Frontiers in molecular biosciences, 10, 1017757-1017757 (2023-03-21)
Genetically-encoded combinatorial peptide libraries are convenient tools to identify peptides to be used as therapeutics, antimicrobials and functional synthetic biology modules. Here, we report the identification and characterization of a cyclic peptide, G4CP2, that interferes with the GAL4 protein, a
Volha A Golubeva et al.
Cells, 9(1) (2020-01-16)
The Zika virus (ZIKV) is a mosquito-borne Flavivirus and can be transmitted through an infected mosquito bite or through human-to-human interaction by sexual activity, blood transfusion, breastfeeding, or perinatal exposure. After the 2015-2016 outbreak in Brazil, a strong link between
Claire E J Moore et al.
Cellular signalling, 28(4), 284-293 (2016-01-23)
Eukaryotic elongation factor 2 kinase (eEF2K) inhibits the elongation stage of protein synthesis by phosphorylating its only known substrate, eEF2. eEF2K is tightly regulated by nutrient-sensitive signalling pathways. For example, it is inhibited by signalling through mammalian target of rapamycin
Hui Tian et al.
Molecular biology of the cell, 32(1), 1-14 (2020-11-12)
Meiotic recombination in most mammals requires recombination hotspot activation through the action of the histone 3 Lys-4 and Lys-36 methyltransferase PRDM9 to ensure successful double-strand-break initiation and repair. Here we show that EWSR1, a protein whose role in meiosis was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service