Skip to Content
Merck
All Photos(1)

Documents

A7340

Sigma-Aldrich

Anti-Glutathione-S-Transferase (GST)–Peroxidase Conjugate antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Glutathione-S-Transferase, Anti-GST

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

peroxidase conjugate

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 27.5 kDa

technique(s)

direct ELISA: 1:10,000
western blot: 1:10,000 using lysates of Escherichia coli induced to express recombinant GST

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Glutathione-S-Transferase (GST) belongs to a family of Phase II detoxification enzymes which are involved in catalyzing the conjugation of glutathione (GSH) with many exogenous and endogenous electrophilic substances. GST is of two kinds- membrane-bound microsomal and cytosolic. The cytosolic GSTs are made of six classes: α, μ, ω, π, θ, and ζ.
The antibody is specific for native as well as denatured-reduced forms of glutathione-S-transferase from Schistosoma japonicum. Anti-GST may be used in various immunoassays to identify the expression of GST fusion proteins.

Specificity

Recombinant target proteins are often expressed as a fusion product with Glutathione-S-Transferase (GST) tags using various expression vector constructs. Thus, antibodies directed against the GST tags of the recombinant constructs can facilitate the purification and study of target proteins.

Immunogen

recombinant GST from Schistosoma japonicum expressed in E. coli.

Application

Anti-Glutathione-S-Transferase (GST)-Peroxidase Conjugate antibody has been used in
  • immunoblotting
  • affinity pull-down assay
  • western blotting
  • enzyme linked immunosorbent assay (ELISA)

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Glutathione-S-Transferase (GST) are involved in protecting cellular macromolecules from the reactive electrophiles. The μ and π transferases regulate the mitogen-activated protein (MAP) kinase pathway. GSTs help in building up cellular resistance against microbial antibiotic, herbicides, insecticides, chemotherapy agents and drugs.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 0.05% MIT.

Preparation Note

Prepared by the two-step glutaraldehyde method described by Avrameas, S., et al., Scand. J. Immunol., 8, Suppl. 7, 7 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flavivirus NS3 and NS5 proteins interaction network: a high-throughput yeast two-hybrid screen
Le B M, et al.
BMC Microbiology, 11(1), 234-234 (2011)
Hiroshi Qadota et al.
Molecular biology of the cell, 27(10), 1606-1620 (2016-03-25)
UNC-89 is a giant polypeptide located at the sarcomeric M-line of Caenorhabditis elegans muscle. The human homologue is obscurin. To understand how UNC-89 is localized and functions, we have been identifying its binding partners. Screening a yeast two-hybrid library revealed
Activating transcription factor 6 derepression mediates neuroprotection in Huntington disease
Naranjo J R, et al.
The Journal of Clinical Investigation, 126(2), 627-638 (2016)
A A Ivanov et al.
Oncogene, 36(42), 5852-5860 (2017-06-20)
Mitogen-activated protein kinase kinase 3 (MKK3) is a dual threonine/tyrosine protein kinase that regulates inflammation, proliferation and apoptosis through specific phosphorylation and activation of the p38 mitogen-activated protein kinase. However, the role of MKK3 beyond p38-signaling remains elusive. Recently, we
G Garrait et al.
Metabolic engineering, 11(3), 148-154 (2009-04-07)
An innovative "biodrug" concept based on oral administration of living recombinant microorganisms as a vehicle to deliver active compounds directly into the digestive tract has recently been developed. To validate this concept, we studied a recombinant Saccharomyces cerevisiae strain in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service