Skip to Content
Merck
All Photos(2)

Documents

165344

Sigma-Aldrich

1-[3-(Dimethylamino)propyl]-3-ethylcarbodiimide methiodide

for peptide synthesis

Synonym(s):

EDC methiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5N=C=N(CH2)3N(CH3)3I
CAS Number:
Molecular Weight:
297.18
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

product name

1-[3-(Dimethylamino)propyl]-3-ethylcarbodiimide methiodide,

form

powder

reaction suitability

reaction type: Coupling Reactions

mp

97-99 °C (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

[I-].CCN=C=NCCC[N+](C)(C)C

InChI

1S/C9H20N3.HI/c1-5-10-9-11-7-6-8-12(2,3)4;/h5-8H2,1-4H3;1H/q+1;/p-1

InChI key

AGSKWMRPXWHSPF-UHFFFAOYSA-M

Application

  • Crosslinking agent

Reactant for:
  • Amide bond forming (amidation) crosslinking reactions
Water-soluble carboxyl modifying reagent for proteins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A K Tsaturyan et al.
Biophysical journal, 77(1), 354-372 (1999-07-02)
Structural changes induced by Joule temperature jumps (T-jumps) in frog muscle fibers were monitored using time-resolved x-ray diffraction. Experiments made use of single, permeabilized fibers that were fully activated after slight cross-linking with 1-ethyl-3-[3-dimethylamino)propyl]carbodiimide to preserve their structural order. After
Heather M Powell et al.
Biomaterials, 28(6), 1084-1092 (2006-11-07)
Collagen-glycosaminoglycan (C-GAG) sponges are commonly utilized as a substitute for the extracellular matrix of dermal tissue. Cultured skin substitutes (CSS) were assessed, after fabrication using sponges cross-linked with 1-ethyl-3-3-dimethylaminopropylcarbodiimide hydrochloride (EDC) at 0, 1, 5, or 50 mm, for development
Takahiro Kishimoto et al.
Biopolymers, 79(3), 163-172 (2005-08-12)
Pro-Hyp-Gly is a characteristic amino acid sequence found in fibrous collagens, and (Pro-Hyp-Gly)(10), which has been widely used as a collagen-model peptide, forms a stable triple-helical structure. Here, we synthesized polypeptides consisting of the Pro-Hyp-Gly sequence by direct poly-condensation of
Cyrille Garnier et al.
Biochemistry, 41(39), 11770-11778 (2002-09-25)
The 90 kDa heat-shock protein (Hsp90) is one of the major stress proteins whose overall structure remains unknown. In this study, we investigated the influence of divalent cations Mg(2+) and Ca(2+) on the hydrodynamic properties and quaternary structure of Hsp90.
T S Taha et al.
Biochemistry, 31(37), 9090-9097 (1992-09-22)
C/57 black mice were immunized with beef heart cytochrome c oxidase, generating 48 hybrid cell lines that secrete antibodies against the different subunits of the enzyme. Immunoblot analysis showed reactions with 7 of the 13 subunits. Among the monoclonal antibodies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service